Skip to Content
Merck
All Photos(1)

Key Documents

TMO362805-0020

Nalgene® Amber Micro Packaging Vial

polypropylene copolymer, vial volume 2.0 mL, self-standing, conical bottom, external thread, untreated, case of 1000 ea

Synonym(s):

light sensitive reagent vial, micro vial

Sign Into View Organizational & Contract Pricing

Select a Size

1000 EACH
€239.00

€239.00


Check Cart for Availability


Select a Size

Change View
1000 EACH
€239.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

€239.00


Check Cart for Availability

material

conical bottom
polypropylene copolymer

feature

self-standing

packaging

case of 1000 ea

manufacturer/tradename

Nalgene 362805-0020

technique(s)

PCR: suitable

vial volume

2.0 mL

Looking for similar products? Visit Product Comparison Guide

General description

Protect light sensitive reagents with Thermo Scientific Nalgene PPCO Amber Micro Packaging Vials. These noncytotoxic and nonpyrogenic vials are molded from high purity, low-metal content polypropylene copolymer resin. They are skirted and feature a conical bottom, allowing recovery of entire contents.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service