Key Documents
TMO140156
Nunc® OmniTray(TM) Single-Well Plate un-notched
polystyrene, sterile, with lid, clear, case of 90 ea
Synonym(s):
microplate, treated microplate
Select a Size
90 EACH
€635.00
€635.00
Estimated to ship on28 April 2025
Select a Size
Change View
90 EACH
€635.00
About This Item
UNSPSC Code:
41122102
NACRES:
NB.13
€635.00
Estimated to ship on28 April 2025
material
clear
polystyrene
sterility
sterile
packaging
case of 90 ea
manufacturer/tradename
Nunc 140156
General description
Made from clear polystyrene with standard microplate footprint, the Thermo Scientific Nunc OmniTrays are ideal for screen libraries and DNA hybridization. Available both as a standard version with 2 notched corners and as a rectangular version optimized for automation. Surface treated for cell culture or non-treated for bacterial growth and colony isolation.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)