Skip to Content
Merck
All Photos(4)

Key Documents

T9075

Sigma-Aldrich

Anti-TRPV4 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-OTRPC4, Anti-Transient Receptor Potential Channel V4

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€832.00

€832.00


Estimated to ship on09 April 2025



Select a Size

Change View
200 μL
€832.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€832.00


Estimated to ship on09 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using ND7/23 (mouse neuroblastoma X rat DRG neuron hybrid cell line) lysate

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

synthetic peptide CDGHQQGYAPKWRAEDAPL corresponding to residues 853-871 of rat TRPV4.

Physical form

Lyophilized from phosphate buffered saline containing 1% BSA and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lyudmyla Borysova et al.
Nature protocols, 16(10), 4650-4675 (2021-08-18)
Ca2+ handling within cardiac myocytes underpins coordinated contractile function within the beating heart. This protocol enables high spatial and temporal Ca2+ imaging of ex vivo multicellular myocardial strips. The endocardial surface is retained, and strips of 150-300-µm thickness are dissected
Ashwini Saxena et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 307(8), R945-R955 (2014-08-01)
Transient receptor potential vanilloid family type 4 (TRPV4) channels are expressed in central neuroendocrine neurons and have been shown to be polymodal in other systems. We previously reported that in the rodent, a model of dilutional hyponatremia associated with hepatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service