Skip to Content
Merck
All Photos(1)

Key Documents

T8786

Sigma-Aldrich

Thr-Phe-Gln-Ala-Tyr-Pro-Leu-Arg-Glu-Ala

≥97% (HPLC)

Synonym(s):

H-Thr-Phe-Gln-Ala-Tyr-Pro-Leu-Arg-Glu-Ala-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H82N14O16
CAS Number:
Molecular Weight:
1195.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C1CCCN1C(=O)C(Cc2ccc(O)cc2)NC(=O)C(C)NC(=O)C(CCC(N)=O)NC(=O)C(Cc3ccccc3)NC(=O)C(N)C(C)O)C(=O)NC(CCCNC(N)=N)C(=O)NC(CCC(O)=O)C(=O)NC(C)C(O)=O

InChI

1S/C55H82N14O16/c1-28(2)25-38(49(79)63-35(13-9-23-60-55(58)59)48(78)64-37(20-22-43(73)74)47(77)62-30(4)54(84)85)66-51(81)41-14-10-24-69(41)53(83)40(27-33-15-17-34(71)18-16-33)68-45(75)29(3)61-46(76)36(19-21-42(56)72)65-50(80)39(26-32-11-7-6-8-12-32)67-52(82)44(57)31(5)70/h6-8,11-12,15-18,28-31,35-41,44,70-71H,9-10,13-14,19-27,57H2,1-5H3,(H2,56,72)(H,61,76)(H,62,77)(H,63,79)(H,64,78)(H,65,80)(H,66,81)(H,67,82)(H,68,75)(H,73,74)(H,84,85)(H4,58,59,60)

InChI key

GNKMMPBLQYIPCI-UHFFFAOYSA-N

Substrates

Retroviral protease substrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kotler et al.
Proceedings of the National Academy of Sciences of the United States of America, 85(12), 4185-4189 (1988-06-01)
Processing of the gag and pol gene precursor proteins of retroviruses is essential for infectivity and is directed by a viral protease that is itself included in one of these precursors. We demonstrate here that small synthetic peptides can be

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service