Skip to Content
Merck
All Photos(1)

Documents

T6039

Sigma-Aldrich

Quartz digital timers

Model T-7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122411
NACRES:
NB.35

size

3 1/2 in. × 1 3/4 in. × 1/2 in.

battery power

1.5 V button cell

Looking for similar products? Visit Product Comparison Guide

General description

Display: 4 digits, 0.5 inch
Special features: 99 minute countdown timer with buzzer at end of countdown; memory recall function; magnet in back of unit

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service