All Photos(1)
About This Item
Linear Formula:
HOC6H4CH2CH(NH2)COOC2H5 · HCl
CAS Number:
Molecular Weight:
245.70
Beilstein:
4725904
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
Assay
≥98% (TLC)
form
powder
color
white to off-white
mp
165 °C
storage temp.
−20°C
SMILES string
Cl.CCOC(=O)[C@@H](N)Cc1ccc(O)cc1
InChI
1S/C11H15NO3.ClH/c1-2-15-11(14)10(12)7-8-3-5-9(13)6-4-8;/h3-6,10,13H,2,7,12H2,1H3;1H/t10-;/m0./s1
InChI key
BQULAXAVRFIAHN-PPHPATTJSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Priya Dey et al.
PloS one, 14(9), e0223216-e0223216 (2019-10-01)
The use of biopolymers as elicitors in controlling plant diseases is gaining momentum world-wide due to their eco-friendly and non-toxic nature. In the present study, we have used an algal biopolymer (sodium alginate) and tested its applicability as an elicitor
A study of the intermolecular interactions of tolmetin/N-acetyl-L-tyrosine ethyl ester complex.
Tito A, Jimenez-Lopez C, et al.
Spectrochimica Acta Part A: Molecular Spectroscopy, 72, 1000-1006 (2009)
C L Penney et al.
Journal of biological standardization, 14(4), 345-349 (1986-10-01)
A simple, rapid, high performance liquid chromatographic (HPLC) method for the analysis of the immunoadjuvant octadecyl tyrosine hydrochloride is described. The HPLC procedure can be applied to the direct determination of amino acid reactants present as contaminants in the adjuvant
Ivo B Rietveld et al.
Journal of pharmaceutical sciences, 100(11), 4774-4782 (2011-06-24)
Polymorphism is important in the field of solid-state behavior of drug molecules because of the continuous drive for complete control over drug properties. By comparing different structures of a series of L-tyrosine alkyl esters, it became apparent that the ethyl
Tatsuro Kijima et al.
Biotechnology letters, 26(8), 655-658 (2004-06-18)
The fluorescence emission wavelength of alpha-chymotrypsin (CT) correlated with its enantioselectivity (E value) for the resolution of DL-tyrosine ethyl ester. The changes in the E value of the CT due to the changes in the solvent composition were closely related
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service