Skip to Content
Merck
All Photos(2)

Key Documents

T4446

SAFC

N-Acetyl-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
223.23
Beilstein:
2697172
EC Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26
Pricing and availability is not currently available.

biological source

non-animal source

Assay

≥98.5%

form

crystalline powder

manufacturer/tradename

Ajinomoto

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, heavy metals, tested

mp

149-152 °C (lit.)

solubility

H2O: 25 mg/mL

application(s)

pharmaceutical (small molecule)

SMILES string

CC(=O)N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C11H13NO4/c1-7(13)12-10(11(15)16)6-8-2-4-9(14)5-3-8/h2-5,10,14H,6H2,1H3,(H,12,13)(H,15,16)/t10-/m0/s1

InChI key

CAHKINHBCWCHCF-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to documentation for this product requires a confidentiality disclosure agreement.

Application

L-Tyrosine is a non-essential amino acid. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. N-Acetyl-L-tyrosine is an acetylated derivative of the essential amino acid L-tyrosine with reported improved nutritional properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hauh-Jyun Candy Chen et al.
Chembiochem : a European journal of chemical biology, 9(7), 1074-1081 (2008-03-21)
Reactive nitrogen species are implicated in inflammatory diseases and cancers. Oxanine (Oxa) is a DNA lesion product originating from the guanine base through exposure to nitric oxide, nitrous acid, or N-nitrosoindoles. Oxanine was found to mediate formation of DNA-protein cross-links
Michael J Napolitano et al.
Chemical research in toxicology, 18(3), 501-508 (2005-03-22)
The reactions of aqueous ClO2 with tyrosine, N-acetyltyrosine, and dopa (3,4-dihydroxyphenylalanine) are investigated from pH 4 to 7. The reaction rates increase greatly with pH to give a series of oxidized products. Tyrosine and N-acetyltyrosine have similar reactivities with second-order
A Rescigno et al.
Biochimica et biophysica acta, 1384(2), 268-276 (1998-07-11)
Tyrosinase is a copper containing protein which catalyzes the hydroxylation of monophenols and the oxidation of diphenols to o-quinones. The monophenolase activity of tyrosinase is characterized by a typical lag time. In this paper the influence of 3-hydroxyanthranilic acid on
T Nishiyama et al.
Journal of biochemistry, 127(3), 427-433 (2000-03-25)
It was previously revealed [Yamaguchi, H., Nishiyama, T., and Uchida, M. (1999) J. Biochem. 126, 261-265] that N-glycans of both the high-mannose and complex types have binding affinity for aromatic amino acid residues. This study shows that free N-glycans protect
Sandrine Perrier et al.
Journal of the American Chemical Society, 131(34), 12458-12465 (2009-08-08)
Steady-state (1)H photo-chemically induced dynamic nuclear polarization (CIDNP) experiments were conducted at 14.1 T on deoxygenated (buffered pH 7) aqueous solutions of [Ru(phen)(3)](2+), [Ru(tap)(2)(phen)](2+), and [Ru(tap)(3)](2+) (tap = 1,4,5,8-tetraazaphenanthrene; phen = 1,10-phenanthroline) in the presence of guanosine-5'-monophosphate or N-acetyltyrosine. For

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service