Skip to Content
Merck
All Photos(3)

Key Documents

T3763

Sigma-Aldrich

α-Thymidine

Synonym(s):

1-(2-Deoxy-α-D-ribofuranosyl)-5-methyluracil

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€153.00
1 G
€795.00

€153.00


Estimated to ship on29 March 2025



Select a Size

Change View
100 MG
€153.00
1 G
€795.00

About This Item

Empirical Formula (Hill Notation):
C10H14N2O5
CAS Number:
Molecular Weight:
242.23
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

€153.00


Estimated to ship on29 March 2025


Assay

≥98% (HPLC)

Quality Level

form

powder

optical activity

[α]25/D 5.0 to 8.0°, c = 2.0% in water

solubility

water: 50 mg/mL, clear, colorless

SMILES string

CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

InChI

1S/C10H14N2O5/c1-5-3-12(10(16)11-9(5)15)8-2-6(14)7(4-13)17-8/h3,6-8,13-14H,2,4H2,1H3,(H,11,15,16)

InChI key

IQFYYKKMVGJFEH-UHFFFAOYSA-N

Application

α-Thymidine (α-Thymidine) is the unnatural epimer of thymidine.

Other Notes

Unnatural epimer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina H Andrade et al.
Journal of chemical information and modeling, 49(4), 1070-1078 (2009-03-20)
Thymidine monophosphate kinase (TMPK) has emerged as an attractive target for developing inhibitors of Mycobacterium tuberculosis growth. In this study the receptor-independent (RI) 4D-QSAR formalism has been used to develop QSAR models and corresponding 3D-pharmacophores for a set of 5'-thiourea-substituted
Y Sato et al.
Nucleic acids symposium series, 44(44), 31-32 (2003-08-09)
In this paper, we report the synthesis of hydroxymethylphosphonate alpha-DNAs and related compounds by use of the H-phosphonate method. These modified alpha-DNAs were designed to improve the inherent poor solubility of well-known methylphosphonate alpha-DNAs by introduction of a more hydrophilic
Sara Van Poecke et al.
Bioorganic & medicinal chemistry, 19(24), 7603-7611 (2011-11-09)
We report on Mycobacterium tuberculosis thymidine monophosphate kinase (TMPKmt) inhibitory activities of a series of new 3'- and 5'-modified thymidine analogues including α- and β-derivatives. In addition, several analogues were synthesized in which the 4-oxygen was replaced by a more

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service