Skip to Content
Merck
All Photos(1)

Key Documents

T2793

Sigma-Aldrich

Nalgene® centrifuge tubes, Oak Ridge Style 3139

nominal capacity 30 mL

Synonym(s):

High-Speed ​​Centrifuge Tubes, Nalgene tubes

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€214.00

€214.00


Check Cart for Availability


Select a Size

Change View
10 EA
€214.00

About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

€214.00


Check Cart for Availability

material

cap (Sealing cap)
polypropylene
polypropylene copolymer
round bottom tube

feature

RCF 50,000 × g

packaging

pack of 10 ea

manufacturer/tradename

Nalgene® 3139-0030

nominal capacity

30 mL

Looking for similar products? Visit Product Comparison Guide

General description

Spin with confidence with when you use these Nalgene® Oak Ridge high-speed PPCO centrifuge tubes complete with sealing caps. These tubes are chemically compatible with a wider variety of laboratory reagents than polycarbonate centrifuge tubes and are repeately autoclavable for extended performance.

Features and Benefits

  • Greater chemical resistance than polycarbonate
  • Autoclavable
  • Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
  • 3139-0050 (T2918) can be used up to 100,605 x g in the T29-8x50 rotor
  • Comes with polypropylene sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
  • Must be filled to 80% of total capacity when centrifuging
  • Autoclavable/Leakproof† when used with supplied Nalgene closure; Repeated autoclaving will reduce the life of PC centrifuge tubes.

Packaging

With sealing cap

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service