Skip to Content
Merck
All Photos(2)

Key Documents

T1575

Sigma-Aldrich

Anti-Phosphotyrosine (pY-256) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect immunofluorescence: 5-10 μg/mL using A431 stimulated by human EGF.
western blot: 0.5-1.0 μg/mL using cell extract of A431 stimulated by human EGF.

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr256)

General description

Protein phosphorylation is a post-translational modification that regulates signal transduction. Phosphorylation of tyrosine residues in proteins has been associated with oncogenesis, cell growth and survival . Phosphotyrosines have also been linked to other cellular functions. For instance, tyrosine 256 phosphorylation in atypical protein kinase C enables its entry into the nucleus of cells . Anti-Phosphotyrosine pTyr256 binds to tyrosine phosphorylated proteins. In immunoblotting, the antibody detects [pTyr256] in random peptides and phospho-L-tyrosine, but does not react with phospho-L-serine, phospho-L-threonine and L-tyrosine.

Immunogen

phospho-L-tyrosine containing random peptide pY-256, conjugated to KLH.

Application

Anti-Phosphotyrosine (pY-256) antibody is suitable for use in western blot (1:1000) . It may also be used in indirect immunofluorescence (5-10 μg/mL using A431 stiμLated by human EGF), immunoblotting and immunocytochemistry.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wendy O White et al.
Journal of cellular biochemistry, 85(1), 42-53 (2002-03-14)
Herein, we employed a combined approach of molecular modeling and site-directed mutagenesis to address the role of tyrosine phosphorylation in transport of atypical protein kinase C (aPKC) into the nucleus. Computer modeling of the three-dimensional structure of the aPKC catalytic
John Rush et al.
Nature biotechnology, 23(1), 94-101 (2004-12-14)
Tyrosine kinases play a prominent role in human cancer, yet the oncogenic signaling pathways driving cell proliferation and survival have been difficult to identify, in part because of the complexity of the pathways and in part because of low cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service