SYNP290170
Synthware™ Crystallizing dish with spout
clear borosilicate glass 3.3, volume 150 mL, diam. × H 80 mm × 40 mm
Synonym(s):
Synthware P290170
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear borosilicate glass 3.3
packaging
pack of 1 ea
manufacturer/tradename
Kemtech P290170
diam. × H
80 mm × 40 mm
volume
150 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Dish, Crystallizing, With Spouts, 80X40mm, 150mL Capacity
Application
kemetch; synthware; dish; crystallizing
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service