Skip to Content
Merck
All Photos(1)

Key Documents

SRP7603

Sigma-Aldrich

DNA Binding Protein-7 (DBP-7), recombinant

recombinant, expressed in E. coli, ≥99% (SDS-PAGE)

Synonym(s):

DNA Binding Protein 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥99% (SDS-PAGE)

form

lyophilized

mol wt

~9.4 kDa

packaging

pkg of 20 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... sso7d(1454006)

General description

DNA Binding Protein-7 (DBP-7) is a N-terminal His-tagged recombinant protein and a member of the Sso7d family of small, abundant, non-specific DNA-binding proteins from the hyperthermophilic Archea Sulfolobus. The 7-kDa protein from Sulfolobus spp. consists of a five stranded, incomplete β-barrel capped at the opening by a C-terminal α-helix; they bind to the minor groove of a DNA duplex via the triple-stranded β-sheet. The topology of the Sulfolobus 7-kDa proteins was found to be similar to that of chromatin organization modifier (chromo) domains and eukaryotic SH3 domains, which are involved in protein–protein interactions. In vitro studies have shown that DBP-7 promotes the annealing of complementary DNA strands, induces negative supercoiling and chaperones the disassembly and renaturation of protein aggregates in an ATP hydrolysis-dependent manner.

Physical form

Sterile filtered and Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. The solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service