Skip to Content
Merck
All Photos(1)

Key Documents

T4GENE32-RO

Roche

T4 Gene 32 Protein

from Escherichia coli(infected with phage T4amN134/amBL292/amE218), solution, optimum pH ~8.0

Synonym(s):

gene 32 protein, t4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€280.00
500 μG
€1,040.00

€280.00


Check Cart for Availability


Select a Size

Change View
100 μG
€280.00
500 μG
€1,040.00

About This Item

UNSPSC Code:
12352202

€280.00


Check Cart for Availability

biological source

Escherichia coli (infected with phage T4amN134/amBL292/amE218)

Quality Level

form

solution

packaging

pkg of 100 μg (10972983001)
pkg of 500 μg (10972991001)

manufacturer/tradename

Roche

optimum pH

~8.0

storage temp.

−20°C

General description

T4 Gene 32 Protein is reported to improve the yield of long PCR products when 0.5 to 1.0 nmol of the protein is included in the reaction. T4 Gene 32 Protein is also reported to increase the yield of PCR products amplified from samples that contain humic acid. The inhibitory effect of humic acid is reduced by a factor of 7 when T4 Gene 32 Protein is included in the PCR (at a concentration of 2.5 μg/100 μl).
The protein is isolated from the triple-mutant T4amN134/ amBL292/ amE 219 defective for the T4 genes 33, 35, and 58 to 61.

Specificity

T4 gene 32 protein is a single-strand specific, helix-destabilizing protein encoded by gene 32 of the phage T4 genome. It is required for the replication of DNA and genetic recombination in Escherichia coli cells that are infected with T4 bacteriophage.[1]

Application

T4 Gene 32 Protein is a single-strand-specific DNA-binding protein. It may be used for:
  • Optimization of PCR (See "Product Description" below.)
  • Stimulation of in vitro DNA synthesis
  • Stabilization of single-stranded regions of DNA or RNA
  • Site-specific mutagenesis experiments (with T4 DNA polymerase and T4 DNA ligase)
  • Helping restriction enzyme digestions go to completion
  • q-PCR[2]

Quality

Absence of contaminants
T4 gene 32 protein is incubated with various nucleic acids that serve as potential nuclease substrates. Results are analyzed by gel electrophoresis. Based on these tests, none of the following are detectable in the preparation according to the current Quality Control procedures:
  • nonspecific exo- and endonucleases
  • single-strand DNases (nicking activities)
  • single-strand-specific exonucleases
  • RNases

Physical form

Protein solution, 1 - 10mg/ml, in storage buffer (20mM Tris-HCl, 100mM NaCl, 1mM EDTA, 0.5mM DTT, 50% glycerol [v/v], pH approximately 8.0)

Preparation Note

Working concentration: 1 to 10 mg/ml

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Bittner et al.
The Journal of biological chemistry, 254(19), 9565-9572 (1979-10-10)
Detailed procedures are presented which allow reproducible preparation of T4 gene 32 protein, a helix-destabilizing protein essential for DNA replication and genetic recombination in T4 bacteriophage-infected Escherichia coli cells. Although 32 protein can be purified to better than 99% homogeneity
Pauline Jacob et al.
International journal of environmental research and public health, 12(3), 2967-2983 (2015-03-13)
A two-year monitoring program of Cryptosporidium parvum oocysts, Giardia duodenalis cysts, Escherichia coli, Clostridium perfringens spores and adenovirus was conducted in three large rivers in France used for recreational activities and as a resource for drinking water production. Fifty-liter river

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service