Skip to Content
Merck
All Photos(1)

Key Documents

SRP6057

Sigma-Aldrich

Activin A from mouse

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

FRP, FSH-releasing protein, Inhibin beta-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

26.2 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | embryo: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... INHBA(3624)

General description

Activin A is a member of the TGF (transforming growth factor)-β family of proteins produced by many cell types throughout development. Mouse Activin A has 100% amino acid sequence identity to human, rat, porcine, bovine and feline proteins. Recombinant mouse Activin A is a nonglycosylated homodimer, containing two 117 amino acid chains, with a total molecular weight of 26.2 kDa.[1][2]

Application

Activin A has been used:
  • to induce differentiation of embryonic stem cells to pancreatic β cells[3]
  • in the induction of bone marrow-derived mesenchymal stem cells to insulin-producing cells[4]
  • to induce endoderm differentiation from human embryonic stem cells[5]
  • as a component of β cell induction medium. [6]

Biochem/physiol Actions

Activins interact with Type I and Type II serine/threonine kinases to signal to SMAD proteins to regulate a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, metabolism, etc. Activin A is a homodimer of two β A chains and is not biologically active until the N terminal propeptide is cleaved from each chain.[1][2][7] Activin A is involved in mesoderm formation and hematopoietic development.

Physical form

Recombinant mouse Activin A is lyophilized with 0.02% TFA.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Centrifuge vial before opening. When reconstituting the product, gently pipet and wash down the sides of the vial to ensure full recovery of the protein into solution. It is recommended to reconstitute the lyophilized product with sterile water, which can then be further diluted to other aqueous solutions.

Other Notes

MGLECDGKVN ICCKKQFFVS FKDIGWNDWI IAPSGYHANY CEGECPSHIA GTSGSSLSFH STVINHYRMR GHSPFANLKS CCVPTKLRPM SMLYYDDGQN IIKKDIQNMI VEECGCS

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inducing embryonic stem cells to differentiate into pancreatic beta cells by a novel three-step approach with activin A and all-trans retinoic acid.
Shi Y
Stem Cells, 23, 656-662 (2005)
Functional analysis of activins during mammalian development.
Matzuk MM
Nature, 374, 354-356 (1995)
Different phenotypes for mice deficient in either activins or activin receptor type II.
Matzuk MM
Nature, 374, 356-360 (1995)
Differentiation of bone marrow-derived mesenchymal stem cells from diabetic patients into insulin-producing cells in vitro.
Sun Y
Chinese Medical Journal (English Edition), 120, 771-776 (2007)
Requirement for activin A and transforming growth factor--beta 1 pro-regions in homodimer assembly.
Gray AM
Science, 247, 1328-1330 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service