Skip to Content
Merck

Skip To

A4941

Activin A human

≥95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, lyophilized powder, suitable for cell culture

Synonym(s):

Activin A Protein, Growth Factor, Human Growth Factor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg
Check Cart for Availability
€ 299,00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
EC Number:
201-069-1
MDL number:
Biological source:
human
Recombinant:
expressed in baculovirus infected insect cells
Assay:
≥95% (SDS-PAGE)
Form:
lyophilized powder
Mol wt:
dimer 26 kDa (containing 116 amino acid residues βA subunits)
Impurities:
endotoxin, tested

€ 299,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Activin A human, ≥95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, lyophilized powder, suitable for cell culture

biological source

human

Quality Segment

recombinant

expressed in baculovirus infected insect cells

assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

1-10 ng/mL ED50

mol wt

dimer 26 kDa (containing 116 amino acid residues βA subunits)

packaging

pkg of 10 μg, pkg of 5 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... INHBA(3624)

General description

Activin A gene with two exons is mapped to human chromosome 2q35. Activin A also known as inhibin α-subunit belongs to the TGF-β (transforming growth factor-β) superfamily of proteins and its expression is exclusive to gonads, placenta and adrenal cortex.

Application

Activin A human has been used as a constituent of modified barth′s saline used in in vitro induction of pronephros.[1] It has also been used as a constituent of Roswell Park memorial institute (RPMI) media to induce pancreatic differentiation of multipotent dermal fibroblasts in vitro.

Biochem/physiol Actions

Activins play a vital role in the regulation of hormonogenesis, cellular homeostasis and differentiation programs.[2] Elevated expression of activin A and B leads to the loss of muscle mass. Both activin A and B have a potential to act as a serum markers of chronic fatigue syndrome/myalgic encephalomyelitis (CFS/ME).

Physical form

Lyophilized from 0.2 μm filtered solution in 10 mM sodium citrate, pH 3.0.

Analysis Note

The biological activity is determined by its ability to inhibit the proliferation of mouse MPC-11 cells.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H8541H8916H4666
biological source

human

biological source

human

biological source

human

biological source

human

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

recombinant

expressed in baculovirus infected insect cells

recombinant

expressed in HEK 293 cells

recombinant

expressed in HEK 293 cells

recombinant

expressed in HEK 293 cells

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

100

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Still not finding the right product?

Explore all of our products under Activin A human


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters