Skip to Content
Merck
All Photos(1)

Key Documents

SRP6001

Sigma-Aldrich

Procathepsin K human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

CTSK, CTSO, CTSO2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

packaging

pkg of 10 μg

concentration

~200 μg/mL

impurities

Endotoxin, tested

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CATK(1513)

General description

Procathepsin K is the inactive proenzyme of cathepsin K. Cathepsin K is a lysosomal cysteine protease and part of the papain cysteine protease family. It is expressed in osteoclasts. The gene encoding this protein is localized on human chromosome 1q21.

Biochem/physiol Actions

Cathepsin K has a role in bone resorption and remodeling. It has a triple helical collagen hydrolase activity. Mutations in the gene encoding this protein have been linked to pycnodysostosis.

Physical form

Liquid (25 mM Tris pH 8.0, 500 mM NaCl)

Other Notes

Procathepsin K can be activated by adjusting the pH 4.0 by adding an equal volume of 100 mM NaAc pH 3.9, 10 mM DTT, 5 mM EDTA followed by incubation for 40 min at RT. Activated mature Cathepsin is highly auto-proteolytic at pH 4.0, and care must be taken to avoid self-proteolysis. If the activated enzyme is not used immediately, we recommend to add methyl methanthiosulfonate (1 mM final concentration; MeS-SO2Me; MMTS) and to freeze the sample in liquid nitrogen or on dry ice. The hydrophobic thiol-reactive compound MMTS modifies cysteine′s by attaching its relatively small, uncharged thiomethyl-blocking group to reactive sulfhydryl groups (Nishimura et al., 1975). This reversible reaction arrests the auto-proteolytic process. The activity of the enzyme can be restored to nearly unmodified levels by adding L-cysteine (3M excess over MMTS) to the enzyme solution.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vito Turk et al.
Biochimica et biophysica acta, 1824(1), 68-88 (2011-10-26)
It is more than 50 years since the lysosome was discovered. Since then its hydrolytic machinery, including proteases and other hydrolases, has been fairly well identified and characterized. Among these are the cysteine cathepsins, members of the family of papain-like
Xianglan Huang et al.
Calcified tissue international, 96(5), 373-378 (2015-03-03)
Pycnodysostosis is a rare autosomal recessive skeletal dysplasia characterized by short stature, osteosclerosis, acro-osteolysis, frequent fractures, and skull deformities. Mutation in the gene encoding cathepsin K (CTSK), which is a lysosomal cysteine protease, has been found to be responsible for
Jaime Toral-López et al.
Journal of investigative medicine : the official publication of the American Federation for Clinical Research, 59(2), 277-280 (2010-11-26)
Pycnodysostosis, an autosomal recessive skeletal dysplasia, is characterized by short stature, osteosclerosis, delayed cranial suture closure, hypoplastic mandible, acro-osteolysis, hypoplastic clavicle, and dental anomalies. The disorder is caused by CTSK gene defects, a gene localized on 1q21. To describe the
Guangxian Zhao et al.
PloS one, 10(8), e0136093-e0136093 (2015-08-25)
Cysteinyl cathepsin K (CatK) is one of the most potent mammalian collagenases involved in cardiovascular disease. Here, we investigated the clinical predictive value of serum CatK levels in patients with chronic heart failure (CHF). We examined 134 patients with CHF
Adeleke H Aguda et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(49), 17474-17479 (2014-11-26)
Cathepsin K is the major collagenolytic protease in bone that facilitates physiological as well as pathological bone degradation. Despite its key role in bone remodeling and for being a highly sought-after drug target for the treatment of osteoporosis, the mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service