Skip to Content
Merck
All Photos(1)

Key Documents

SML2259

Sigma-Aldrich

Lapatinib

≥98% (HPLC)

Synonym(s):

Lapatinib, GD 2016, GSK 572016, GW-572016, GW572016, N-(3-Chloro-4-((3-fluorophenyl)methoxy)phenyl)-6-(5-(((2-(methylsulfonyl)ethyl)amino)methyl)-2-furanyl)-4-quinazolinamine, N-(3-Chloro-4-((3-fluorophenyl)methoxy)phenyl)-6-(5-((2-methylsulfonylethylamino)methyl)-2-furyl)quinazolin-4-amine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€54.30
50 MG
€219.00

€54.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€54.30
50 MG
€219.00

About This Item

Empirical Formula (Hill Notation):
C29H26ClFN4O4S
CAS Number:
Molecular Weight:
581.06
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€54.30


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CS(=O)(CCNCC1=CC=C(C2=CC=C3N=CN=C(C3=C2)NC4=CC=C(C(Cl)=C4)OCC5=CC=CC(F)=C5)O1)=O

InChI

1S/C29H26ClFN4O4S/c1-40(36,37)12-11-32-16-23-7-10-27(39-23)20-5-8-26-24(14-20)29(34-18-33-26)35-22-6-9-28(25(30)15-22)38-17-19-3-2-4-21(31)13-19/h2-10,13-15,18,32H,11-12,16-17H2,1H3,(H,33,34,35)

InChI key

BCFGMOOMADDAQU-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lapatinib acts as a dual tyrosine kinase inhibitor of epidermal growth factor receptor (EGFR/ErbB1) and human epidermal growth factor receptor 2 (HER2/ErbB2). It competes for its binding site on the tyrosine kinase domain with adenosine triphosphate (ATP). Lapatinib mediates has been studied in vitro in the inhibition of cell growth as well as and apoptosis induction in several human cancer cells such as breast, lung, vulva, gastric, and head and neck cancer.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Lact.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongwei Zhang et al.
Molecular cancer therapeutics, 7(7), 1846-1850 (2008-07-23)
Epidermal growth factor receptor (EGFR/ErbB1) and HER2 (ErbB2/neu), members of the ErbB receptor tyrosine kinase family, are frequently overexpressed in breast cancer and are known to drive tumor growth and progression, making them promising targets for cancer therapy. Lapatinib is
Lapatinib in breast cancer
Bilancia D, et al.
Annals of Oncology, 18, vi26-vi30 (2007)
Richard M Tehan et al.
Journal of natural products, 85(5), 1363-1373 (2022-05-03)
The genome of entomopathogenic fungus Tolypocladium inflatum Gams encodes 43 putative biosynthetic gene clusters for specialized metabolites, although genotype-phenotype linkages have been reported only for the cyclosporins and fumonisins. T. inflatum was cultured in defined minimal media, supplemented with or
Sharad Verma et al.
PloS one, 13(2), e0190942-e0190942 (2018-02-02)
HER-2 belongs to the human epidermal growth factor receptor (HER) family. Via different signal transduction pathways, HER-2 regulates normal cell proliferation, survival, and differentiation. Recently, it was reported that MCF10A, BT474, and MDA-MB-231 cells bearing the HER2 K753E mutation were
Egle Avizienyte et al.
The Biochemical journal, 415(2), 197-206 (2008-07-01)
Recent clinical data indicates that the emergence of mutant drug-resistant kinase alleles may be particularly relevant for targeted kinase inhibitors. In order to explore how different classes of targeted therapies impact upon resistance mutations, we performed EGFR (epidermal-growth-factor receptor) resistance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service