Skip to Content
Merck
All Photos(1)

Key Documents

SRP5192

Sigma-Aldrich

IRS1 (600-1245), GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HIRS-1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€326.00

€326.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
€326.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

€326.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~118 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IRS1(3667)

General description

IRS1 is the substrate for the insulin tyrosine kinase receptor and is found in a variety of insulin-responsive cells and tissues. IRS1 protein has no intrinsic enzymatic activity but acts as a docking protein, via the SH2 domains, for mediating the insulin downstream signaling events. IRS1 has been shown to associate with the 14-3-3 family of proteins and this could play a role in the regulation of insulin sensitivity by interrupting the association between the insulin receptor and IRS1. IRS1 may be associated with colorectal cancer and diet and related factors may affect the risk by modifying plasma insulin levels. Thus, the inter-individual variation in insulin signaling mediated by IRS1 may play a plausible role in the development of colorectal cancer.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Ogihara et al.
The Journal of biological chemistry, 272(40), 25267-25274 (1997-10-06)
Insulin binding to its receptor induces the phosphorylation of cytosolic substrates, insulin receptor substrate (IRS)-1 and IRS-2, which associate with several Src homology-2 domain-containing proteins. To identify unique IRS-1-binding proteins, we screened a human heart cDNA library with 32P-labeled recombinant
M L Slattery et al.
Breast cancer research and treatment, 104(2), 197-209 (2006-10-20)
An insulin-related pathway to breast cancer has been hypothesized. We examine the 19 CA repeat of the IGF1 gene, the -202 C > A IGFBP3, the G972R IRS, and the G1057D IRS2 polymorphisms among 1,175 non-Hispanic white (NHW) and 576

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service