Skip to Content
Merck
All Photos(1)

Key Documents

SRP5137

Sigma-Aldrich

STAT1 β, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp686B04100, ISGF-3, STAT91

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€236.00

€236.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
€236.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€236.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~118 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... STAT1(6772)

General description

STAT1β is a member of the signal transducers and activators of transcription (STAT) family of proteins that carry out a dual function: signal transduction and activation of transcription. STAT1β transcription factor is specific for the IFN pathway and plays a central role in mediating many, if not all, IFN-dependent biological responses. Presence of STAT1β leads to an efficient antiviral response when cells were infected with virus suggesting that a STAT-dependent pathway is activated following virus infection by endogenously produced IFN. Virus-induced STAT protein translocation from the cytoplasmic compartment can be detected within 3 h of infection.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Improta et al.
Cytokine, 9(6), 383-393 (1997-06-01)
Interferon (IFN) signalling appears to be the predominant pathway that leads to the development of the host defence against virus. However, other mechanisms that are IFN independent may also be involved. The Stat1 transcription factor is specific for the IFN

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service