Skip to Content
Merck

Skip To

C4237

Protamine Kinase, Cytosolic from bovine kidney

>90% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

Cytosolic Protamine Kinase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 vial
Check Cart for Availability
€ 802,00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
MDL number:
Specific activity:
≥15,000 units/mg protein
Assay:
>90% (SDS-PAGE)

€ 802,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

>90% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

≥15,000 units/mg protein

mol wt

45 kDa

packaging

vial of 100 ng

shipped in

dry ice

storage temp.

−70°C

Application

Cytosolic Protamine Kinase (cPK) is isolated from bovine kidney and has a molecular mass of approximately 45 kDa. It phosphorylates the eukaryotic Initiation Factor 4E (eIF4E), which initiates translation by mRNA. It is used to study protein synthesis and various cellular processes.

Biochem/physiol Actions

Cytosolic protamine kinase is involved in the regulation of protein synthesis and is indirectly associated with numerous cellular processes. Cytosolic protamine kinase is a distinct insulin-stimulated kinase involved in the phosphorylation of eukaryotic Initiation Factor 4E (eIF4E) which is key to initiating translation by mRNA. This protein appears to be inactivated by protein phosphatase 2A family members and may also be inhibited by microcystin, okadeic acid, and ATP. The phosphorylation process is reversible and MBPK1 and MBPK2 (Myelin Basic Protein Kinase 1 and 2) may reactivate cytosolic protamine kinase [1].

Physical form

Solution in 50 μl of 50 mM Tris-HCl, pH 7.0, containing 14 mM β-mercaptoethanol, 1mM benzamidine, 0.1 mM PMSF, 1mM EDTA, 50% glycerol

Other Notes

One unit will incorporate 1 nmol of phosphate using protamine sulfate as a substrate per minute at pH 7.0 at 30 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A8850M1816M1941
specific activity

≥15,000 units/mg protein

specific activity

~17,000 units/mg protein

specific activity

≥300 units/mg protein

specific activity

≥1000 units/mg protein

assay

>90% (SDS-PAGE)

assay

-

assay

>90% (SDS-PAGE)

assay

>90% (SDS-PAGE)

form

buffered aqueous glycerol solution

form

aqueous solution

form

solution

form

solution

mol wt

45 kDa

mol wt

36 kDa

mol wt

40 kDa

mol wt

40 kDa

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters