Skip to Content
Merck
All Photos(1)

Key Documents

SRP3299

Sigma-Aldrich

GDF-5 mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

BMP-14, CDMP-1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€350.00

€350.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
€350.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€350.00


Usually ships in 2 weeks.

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

0.08-0.12 μg/mL

mol wt

27.0 kDa

packaging

pkg of 50 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... GDF5(14563)

General description

Mutations in the GDF-5 gene have been implicated in Grebe Syndrome, which is characterized by short stature, extra digits, short and deformed extremities, and in Hunter-Thompson type dwarfism. The mature and functional form of GDF-5 is a homodimer of two 120 amino-acid polypeptide chain (monomers) linked by a single disulfide bond. Each GDF-5 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 27 amino-acid signal peptide and a 348 amino-acid propeptide. This precursor undergoes intracellular dimerization, and upon secretion it is processed by a furin-type protease.

Biochem/physiol Actions

GDF-5 is expressed in long bones during embryonic development and postnatally in articular cartilage. Recombinant murine GDF-5 is a 27.0 kDa homodimeric disulfide-linked protein consisting of two 120 amino acids.

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eiichi Hinoi et al.
Biochemical and biophysical research communications, 450(1), 255-260 (2014-06-20)
We have previously demonstrated promotion by growth differentiation factor-5 (GDF5) of brown adipogenesis for systemic energy expenditure through a mechanism relevant to activating the bone morphological protein (BMP) receptor/mothers against decapentaplegic homolog (Smad)/peroxisome proliferator-activated receptor gamma co-activator 1α (PGC-1α) pathway.
Cartilage-derived morphogenetic proteins. New members of the transforming growth factor-beta superfamily predominantly expressed in long bones during human embryonic development.
Chang, C.H. et al.
The Journal of Biological Chemistry, 45, 28227-28234 (1994)
Zhou Pei et al.
Archives of biochemistry and biophysics, 560, 27-35 (2014-08-01)
Adipocyte differentiation is key to determining the number of adipocytes during the development of obesity. Recent studies have shown that growth differentiation factor-5 (GDF5) promotes brown adipogenesis, however its role in white adipogenesis is still uncertain. The aim of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service