Skip to Content
Merck

Skip To

M2435

c-Myc Peptide

≥97%, lyophilized powder

Synonym(s):

c-Myc tag

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 108,00
4 mg
Check Cart for Availability
€ 294,00
25 mg
Check Cart for Availability
€ 1.370,00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.56
MDL number:

€ 108,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

c-Myc Peptide, lyophilized powder

biological source

synthetic

Quality Segment

assay

≥97%

form

lyophilized powder

mol wt

1203.3 Da

concentration

5-10 μg/mL (Suggested working concentration to compete out the binding of c-Myc)

technique(s)

protein extraction: suitable

storage temp.

−20°C

General description

The c-Myc Peptide is a synthetic peptide with an amino acid sequence that corresponds to the amino acids 410-419 of the C-terminal of human c-myc.

Application

Useful for displacement of c-Myc-tagged fusion proteins bound to anti-c-Myc antibodies in immunoassays. The successful inhibition of antibody binding by c-Myc peptide demonstrates binding is specific.

Preparation Note

Dissolve in water to a final concentration of 5 mg/mL.

Other Notes

Amino acid sequence is corresponds to amino acids 410-419 of the C-terminal of human c-Myc.
Lyophilized from 0.1% TFA in H2O

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V7754SAE0201SAB4200742
technique(s)

protein extraction: suitable

technique(s)

-

technique(s)

-

technique(s)

immunoblotting: 1:250-1:500 using lysate of HEK-293T cells over expressing c-Myc fusion protein, immunohistochemistry: suitable

biological source

synthetic

biological source

-

biological source

-

biological source

mouse

assay

≥97%

assay

≥97% (HPLC)

assay

-

assay

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

suspension

form

lyophilized powder

mol wt

1203.3 Da

mol wt

calculated mol wt 1524.78 Da

mol wt

-

mol wt

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters