Skip to Content
Merck
All Photos(1)

Key Documents

SRP0303

Sigma-Aldrich

HDAC Class 2a Substrate, fluorogenic

≥95% (HPLC)

Synonym(s):

HDAC, histone deacetylase

Sign Into View Organizational & Contract Pricing

Select a Size

250 NMOL
€281.00

€281.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
250 NMOL
€281.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€281.00


Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (HPLC)

form

aqueous solution

packaging

pkg of 250 nmol

shipped in

dry ice

storage temp.

−70°C

General description

A fluorogenic, acetylated peptide substrate for HDAC4, 5, 7 and 9.

Application

Study enzyme kinetics, and screen small molecule inhibitors of HDAC class2a for drug discovery and HTS applications.

Physical form

Supplied as a 5 mM solution in DMSO.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service