Skip to Content
Merck
All Photos(1)

Key Documents

SRP0290

Sigma-Aldrich

Cathepsin F Active human

recombinant, expressed in FreeStyle 293-F cells, ≥90% (SDS-PAGE)

Synonym(s):

CATSF, CLN13

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€599.00

€599.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
€599.00

About This Item

EC Number:
UNSPSC Code:
12352204
NACRES:
NA.54

€599.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in FreeStyle 293-F cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

specific activity

≥18 pmol/min-μg

mol wt

53 kDa

technique(s)

activity assay: suitable

solubility

soluble
water: soluble

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CTSF(8722)

General description

Research area: CELL SIGNALING

Human cathepsin F (GenBank Accession No. NM_003793), CD33 signal peptide(amino acids 1-16) + cathepsin S (amino acids 20-end), with C-terminal HIS tag, MW = 53 kDa (more than 58 kDa after post-modification), expressed in FreeStyle 293-F cells.

Application

Active human cathepsin F is useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling. Active human cathepsin F has been used in a study to assess its implications for the development of novel immunomodulators. [1] Active human cathepsin F has also been used in a study to investigate microarray expression analysis and identification of serum biomarkers for Niemann-Pick disease. [2]

Biochem/physiol Actions

Cathepsin F (CtsF) are endopeptidases that play a role in lysosomal protein degradation. Beyond its involvement in protein recycling, CtsF has also been associated with immune responses mediated by Toll-like receptors (TLRs) and antigen processing.[3] Additionally, mutation in the cathepsin F gene is related to the development of familial papillary thyroid cancer [4], type B Kufs disease, defective lysosomal degradation and abnormal autophagy, and Alzheimer′s disease.[5]

Unit Definition

One unit is defined as the amount of enzyme that will cleave 1 pmol of substrate per min at 37°C

Physical form

Formulated in 45 mM sodium acetate, 135 mM NaCl and 10% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Legal Information

FreeStyle is a trademark of Invitrogen Corp.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cathepsins in the Pathophysiology of Mucopolysaccharidoses: New Perspectives for Therapy
Pasquale VD, et al.
Cells, 9(4), 979-979 (2020)
Celine V M Cluzeau et al.
Human molecular genetics, 21(16), 3632-3646 (2012-05-24)
Niemann-Pick disease type C (NPC) is a lysosomal storage disorder characterized by liver disease and progressive neurodegeneration. Deficiency of either NPC1 or NPC2 leads to the accumulation of cholesterol and glycosphingolipids in late endosomes and early lysosomes. In order to
Cathepsin F genetic mutation is associated with familial papillary thyroid cancer
Wang Y, et al.
Journal of Separation Science, 364(4), 414-424 (2022)
Salma Teimoori et al.
Parasitology international, 66(4), 443-447 (2016-05-04)
Diagnosis of Opisthorchis viverrini infection by conventional stool examination is increasingly difficult due to the low intensity of the infection after several rounds of control programmes in endemic regions as well as coinfections with intestinal flukes. Therefore sensitive and specific
The Crystal Structure of Human Cathepsin F and Its Implications for the Development of Novel Immunomodulators
Somoza JR, Palmer JT, Ho JD.
Journal of Molecular Biology, 322(3), 559-568 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service