Skip to Content
Merck
All Photos(2)

Key Documents

T2605

Sigma-Aldrich

StableCell Trypsin Solution

5X, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red

Synonym(s):

Trypsin-EDTA solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€20.40
500 ML
€68.20

€20.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€20.40
500 ML
€68.20

About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.78

€20.40


Check Cart for Availability

Request a Bulk Order

Quality Level

sterility

sterile-filtered

product line

BioReagent

concentration

5X

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

The typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.

Biochem/physiol Actions

Trypsin cleaves peptides on the C-terminal side of lysine and arginine residues. The rate of hydrolysis of this reaction is slowed if an acidic residue is on either side of the cleavage site and hydrolysis is stopped if a proline residue is on the carboxyl side of the cleavage site. The optimal pH for trypsin activity is 7-9. Trypsin can also act to cleave ester and amide linkages of synthetic derivatives of amino acids. EDTA is added to trypsin solutions as a chelating agent that neutralizes calcium and magnesium ions that obscure the peptide bonds on which trypsin acts. Removing these ions increases the enzymatic activity.

Features and Benefits

StableCell trypsin is formulated as a gentle solution for cell detachment. Enzyme activity is retained when stored at 2-8°C, and our studies even show >90% activity is retained when left at room temperatures for up to 7 weeks!
  • Save Time - no more freeze or thaw cycles
  • Save Space - store at 2-8°C and free up your freezer
  • Does not need to be aliquoted
  • Only the best for your cells - manufactured in GMP environment

Components

Trypsin consists of a single chain polypeptide of 223 amino acid residues, produced by the removal of the N-terminal hexapeptide from trypsinogen which is cleaved at the Lys - lle peptide bond. The sequence of amino acids is cross-linked by 6 disulfide bridges. This is the native form of trypsin, beta-trypsin. BETA-trypsin can be autolyzed, cleaving at the Lys - Ser residue, to produce alpha-trypsin. Trypsin is a member of the serine protease family.

Preparation Note

Incubating cells with too high a trypsin concentration for a long period can damage cell membranes and kill the cells. Solubilizing trypsin or diluting it from a concentrated solution should be done with a buffered salt solution containing no Ca2+ or Mg2+.

Storage and Stability

Recommended storage is 2-8°C upon arrival. During stability studies, data showed that this product retains ≥90% of its activity when stored at 37°C for up to 8 weeks.

Legal Information

StableCell is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles B Trelford et al.
Biology open, 9(11) (2020-11-11)
Transforming growth factor beta (TGFβ) promotes tumorigenesis by suppressing immune surveillance and inducing epithelial to mesenchymal transition (EMT). TGFβ may augment tumorigenesis by activating autophagy, which protects cancer cells from chemotherapy and promotes invasive and anti-apoptotic properties. Here, we assess

Protocols

StableCell™ Trypsin solutions are designed to perform cell detachment as standard trypsin solutions do, without the need to aliquot, freeze, and thaw. This saves significant time before passaging.

Related Content

Discover our primary human intrahepatic biliary epithelial cells, also known as cholangiocytes. Explore our plating and maintenance media formulations and protocol for culture.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service