Skip to Content
Merck
All Photos(3)

Key Documents

SRE0046

Sigma-Aldrich

Sodium deoxycholate

Suitable for manufacturing of diagnostic kits and reagents

Synonym(s):

3α,12α-Dihydroxy-5β-cholanic acid sodium salt, 7-Deoxycholic acid sodium salt, Desoxycholic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€267.00
500 G
€623.00

€267.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€267.00
500 G
€623.00

About This Item

Empirical Formula (Hill Notation):
C24H39NaO4
CAS Number:
Molecular Weight:
414.55
Beilstein:
3581950
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

€267.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

mol wt

micellar avg mol wt 1200-5000

aggregation number

3-12

technique(s)

DNA purification: suitable
RNA purification: suitable
electrophoresis: suitable

CMC

2-6 mM (20-25°C)

HLB

16

application(s)

diagnostic assay manufacturing

storage temp.

room temp

SMILES string

[Na+].[H][C@]12CC[C@@]3([H])[C@]4([H])CC[C@H]([C@H](C)CCC([O-])=O)[C@@]4(C)[C@@H](O)C[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C24H40O4.Na/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3;/h14-21,25-26H,4-13H2,1-3H3,(H,27,28);/q;+1/p-1/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-;/m1./s1

InChI key

FHHPUSMSKHSNKW-SMOYURAASA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Comparison of SPEED, S-Trap, and In-Solution-Based Sample Preparation Methods for Mass Spectrometry in Kidney Tissue and Plasma.: This research compares different sample preparation methods for mass spectrometry, highlighting the efficacy of sodium deoxycholate in improving protein extraction efficiency (Templeton EM et al., 2023).

  • SimPLIT: Simplified Sample Preparation for Large-Scale Isobaric Tagging Proteomics.: This research introduces a simplified sample preparation method for large-scale proteomics, leveraging sodium deoxycholate to improve peptide and protein identification rates (Sialana FJ et al., 2022).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service