Skip to Content
Merck
All Photos(1)

Documents

SML3870

Sigma-Aldrich

Fuzapladib

≥98% (HPLC)

Synonym(s):

IS 741, IS-741, N-(2-((ethylsulfonyl)amino)-5-(trifluoromethyl)-3-pyridinyl)-3-yl]cyclohexanecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20F3N3O3S
CAS Number:
Molecular Weight:
379.40
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Fuzapladib (IS-741) is a cell penetrant and potent phospholipase A2 (PLA2) inhibitor. Fuzapladib decelerate the progression of pancreatic fibrosis through monocyte infiltration in dogs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of IS-741 on ethionine-induced acute pancreatitis in rats: relation to pancreatic acinar cell regeneration
Journal of Gastroenterology, 38(3), 260-267 (2003)
Nanocrystal solid dispersion of fuzapladib free acid with improved oral bioavailability
Biopharmaceutics & Drug Disposition, 43(3), 89-97 (2022)
Effects of IS-741, a synthetic anti-inflammatory agent, on bleomycin-induced lung injury in mice
Lung, 187(5), 331-339 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service