Skip to Content
Merck
All Photos(1)

Key Documents

SML3629

Sigma-Aldrich

HS-276

≥98% (HPLC)

Synonym(s):

HS 276, HS276, N1-[6-(1-Piperidinylmethyl)-1-propyl-1H-benzimidazol-2-yl]-1,3-benzenedicarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29N5O2
CAS Number:
Molecular Weight:
419.52
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CC(C(NC2=NC3=C(N2CCC)C=C(CN4CCCCC4)C=C3)=O)=C1)N

InChI key

NPEMHKSMWPZEOV-UHFFFAOYSA-N

Biochem/physiol Actions

HS-276 is an orally active, potent and selective TGF-β-activated kinase-1 (TAK-1; TAK1) inhibitor (Ki = 2.5 nM; IC50 = 8.25 nM) that significantly reduced cytokines production in THP-1 human macrophages upon LPS/IFNγ treatment (IC50 = 138 nM/TNF, 201 nM/IL-6, and 234 nM/IL-1β). HS-276 effectively attenuates arthritic-like symptoms in the CIA mouse rheumatoid arthritis (RA) model (10-30 mg/kg/d p.o. or 25 mg/kg/d i.p.).
Orally available, potent and selective TGF-β-activated kinase-1 (TAK-1; TAK1) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott Scarneo et al.
ACS chemical biology, 17(3), 536-544 (2022-03-03)
Selective targeting of TNF in inflammatory diseases such as rheumatoid arthritis (RA) has provided great therapeutic benefit to many patients with chronic RA. Although these therapies show initially high response rates, their therapeutic benefit is limited over the lifetime of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service