Skip to Content
Merck
All Photos(1)

Key Documents

SML3314

Sigma-Aldrich

PF-00835231

≥98% (HPLC)

Synonym(s):

; N-((S)-1-((S)-4-Hydroxy-3-oxo-1-((S)-2-oxopyrrolidin-3-yl)butan-2-ylamino)-4-methyl-1-oxopentan-2-yl)-4-methoxy-1H-indole-2-carboxamide, N-[(1S)-1-[[[(1S)-3-Hydroxy-2-oxo-1-[[(3S)-2-oxo-3-pyrrolidinyl]methyl]propyl]amino]carbonyl]-3-methylbutyl]-4-methoxy-1H-indole-2-carboxamide, PF 00835231, PF 835231, PF-835231, PF00835231, PF835231

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32N4O6
CAS Number:
Molecular Weight:
472.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

COC1=C2C(NC(C(N[C@@H](CC(C)C)C(N[C@H](C(CO)=O)C[C@H]3C(NCC3)=O)=O)=O)=C2)=CC=C1

InChI

1S/C24H32N4O6/c1-13(2)9-18(23(32)27-17(20(30)12-29)10-14-7-8-25-22(14)31)28-24(33)19-11-15-16(26-19)5-4-6-21(15)34-3/h4-6,11,13-14,17-18,26,29H,7-10,12H2,1-3H3,(H,25,31)(H,27,32)(H,28,33)/t14-,17-,18-/m0/s1

InChI key

QDIMHKWNHMVDJB-WBAXXEDZSA-N

Biochem/physiol Actions

Potent and selective corona virus hCoV, SARS CoV-1 & CoV-2 main protease (Mpro; 3CLpro) substrate site Cys145 covalent inhibitor with low host cytotoxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert L Hoffman et al.
Journal of medicinal chemistry, 63(21), 12725-12747 (2020-10-16)
The novel coronavirus disease COVID-19 that emerged in 2019 is caused by the virus SARS CoV-2 and named for its close genetic similarity to SARS CoV-1 that caused severe acute respiratory syndrome (SARS) in 2002. Both SARS coronavirus genomes encode

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service