Skip to Content
Merck
All Photos(1)

Key Documents

SML2320

Sigma-Aldrich

Ornipressin acetate salt

≥98% (HPLC)

Synonym(s):

8-L-Ornithinevasopressin acetate, Cys-Tyr-Phe-Gln-Asn-Cyes-Pro-Orn-Gly-NH2 [Disulfide Bridge: 1-6] acetate, Onithine-vasopressin acetate, Orn8-vasopressin acetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€110.00
25 MG
€441.00

€110.00


Estimated to ship on28 May 2025



Select a Size

Change View
5 MG
€110.00
25 MG
€441.00

About This Item

Empirical Formula (Hill Notation):
C45H63N13O12S2 · xC2H4O2
CAS Number:
Molecular Weight:
1042.19 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€110.00


Estimated to ship on28 May 2025


Assay

≥98% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

S1SC[C@@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@@H](C1)C(=O)N4[C@@H](CCC4)C(=O)N[C@@H](CCCN)C(=O)NCC(=O)N)CC(=O)N)CCC(=O)N)Cc3ccccc3)Cc2ccc(cc2)O)N.OC(=O)C

InChI key

AYJSELDHDXYDKX-HPILINOVSA-N

Biochem/physiol Actions

Ornipressin is a potent vasoconstrictor agent used in surgery for hemostasis and in regional anesthesia for prolongation of nerve block. Ornipressin is a vasopressin agonist specific for the V1 receptor with low antidiuretic activity.
vasopressin agonist specific for the V1 receptor with low antidiuretic activity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sammy Ngichabe et al.
Annals of surgical innovation and research, 9, 10-10 (2015-11-17)
Myomectomy is a surgical technique used for removal of uterine fibroids and historically hysterectomy has represented the mainstay of treatment. The options of conservative surgical approaches mainly aim at retention of fertility but have to be balanced against potential risks
Claudia Tüller et al.
Respiration; international review of thoracic diseases, 71(4), 397-401 (2004-08-19)
Ornipressin and terlipressin, two ADH-derivates, are instilled endobronchially for bronchoscopy- related bleeding without sufficient evidence of efficacy, nor safety. To compare the immediate hemodynamic effects of ornipressin and terlipressin used for procedure-related bleeding during flexible bronchoscopy. This retrospective study included
M Guevara et al.
Hepatology (Baltimore, Md.), 27(1), 35-41 (1998-01-13)
Hepatorenal syndrome is caused by a marked vasoconstriction of the renal circulation. It is suggested that the renal vasoconstriction is related to an overactivity of vasoconstrictor systems secondary to a vasodilation of the arterial circulation that causes a reduction in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service