Skip to Content
Merck
All Photos(1)

Key Documents

SML3159

Sigma-Aldrich

WM-2474

≥98% (HPLC)

Synonym(s):

WM-2474, MOZ-IN-2, N′-(2-Fluoro-5-(pyridazin-4-yl)benzoyl)benzenesulfonohydrazide, N′-(Benzenesulfonyl)-2-fluoro-5-pyridazin-4-yl-benzohydrazide, WM 1119 inactive control, WM 1119 negative control, WM 2474, WM-1119 inactive control, WM-1119 negative control, WM1119 inactive control, WM1119 negative control, WM2474

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€62.80
25 MG
€272.00

€62.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€62.80
25 MG
€272.00

About This Item

Empirical Formula (Hill Notation):
C17H13FN4O3S
CAS Number:
Molecular Weight:
372.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€62.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=C(F)C=CC(C2=CC=NN=C2)=C1)NNS(=O)(C3=CC=CC=C3)=O

InChI

1S/C17H13FN4O3S/c18-16-7-6-12(13-8-9-19-20-11-13)10-15(16)17(23)21-22-26(24,25)14-4-2-1-3-5-14/h1-11,22H,(H,21,23)

InChI key

WYMCVPPNOFFNGE-UHFFFAOYSA-N

Biochem/physiol Actions

Inactive analog of and negative control for the potent and selective, MYST family histone acetyltransferases KAT6A and KAT6B inhibitor WM-1119.
WM-2474 is an inactive analog of and negative control (KAT6B IC50 >125 μM) for the potent and selective, MYST family histone acetyltransferases KAT6A and KAT6B inhibitor WM-1119.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel L Priebbenow et al.
Journal of medicinal chemistry, 63(9), 4655-4684 (2020-03-03)
A high-throughput screen designed to discover new inhibitors of histone acetyltransferase KAT6A uncovered CTX-0124143 (1), a unique aryl acylsulfonohydrazide with an IC50 of 1.0 μM. Using this acylsulfonohydrazide as a template, we herein disclose the results of our extensive structure-activity
Jonathan B Baell et al.
Nature, 560(7717), 253-257 (2018-08-03)
Acetylation of histones by lysine acetyltransferases (KATs) is essential for chromatin organization and function1. Among the genes coding for the MYST family of KATs (KAT5-KAT8) are the oncogenes KAT6A (also known as MOZ) and KAT6B (also known as MORF and
Laura MacPherson et al.
Nature, 577(7789), 266-270 (2019-12-13)
Acute myeloid leukaemia (AML) is a heterogeneous disease characterized by transcriptional dysregulation that results in a block in differentiation and increased malignant self-renewal. Various epigenetic therapies aimed at reversing these hallmarks of AML have progressed into clinical trials, but most

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service