Skip to Content
Merck
All Photos(1)

Key Documents

SML3112

Sigma-Aldrich

cis-22a

≥98% (HPLC)

Synonym(s):

1-[cis-4-(3-Methylphenyl)cyclohexyl]-4-(3-pyridinyl)piperazine, trifluoroacetate (1:2), cis-1-(4-(3-Methylphenyl)cyclohexyl)-4-((pyridin-4-yl)methyl)piperazine, trifluoroacetate (1:2)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€94.90
25 MG
€381.00

€94.90


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€94.90
25 MG
€381.00

About This Item

Empirical Formula (Hill Notation):
C22H29N3 · 2CF3CO2H
CAS Number:
Molecular Weight:
563.53
UNSPSC Code:
12352200
NACRES:
NA.77

€94.90


Estimated to ship on24 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC=CC([C@H]2CC[C@@H](N3CCN(C4=CN=CC=C4)CC3)CC2)=C1.O=C(O)C(F)(F)F.O=C(O)C(F)(F)F

InChI key

UMKPISXYUQWXJJ-XWOHEVPJSA-N

Related Categories

Biochem/physiol Actions

Potent and highly selective TrpV6 (CaT1, ECaC2) inhibitor that selectively inhibits the growth of TrpV6-expressing, but not TrpV6-negative cancer cultures.
cis-22a is a potent and highly selective TrpV6 (CaT1, ECaC2) inhibitor (Cd2+ influx IC50 = 320 nM; hTRPV6 transfected HEK293) with much reduced and little potency against other calcium channels and related TRP targets. cis-22a selectively inhibits the growth of TrpV6-expressing breast cancer T47D (IC50 = 25 μM), but not TrpV6-negative ovarian carcinoma SKOV3 cultures (IC50 >50 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service