Skip to Content
Merck
All Photos(1)

Key Documents

SML1757

Sigma-Aldrich

GSK864

≥98% (HPLC)

Synonym(s):

(S)-1-(4-Fluorobenzyl)-N3-(4-methoxy-3,5-dimethylphenyl)-7-methyl-5-(1H-pyrrole-2-carbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3,7-dicarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€316.00

€316.00


Check Cart for Availability


Select a Size

Change View
5 MG
€316.00

About This Item

Empirical Formula (Hill Notation):
C30H31FN6O4
CAS Number:
Molecular Weight:
558.60
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€316.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -66 to -76°, c = 0.5 in methanol

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CN1)N2C[C@@](C(N)=O)(C)C(N(CC3=CC=C(F)C=C3)N=C4C(NC5=CC(C)=C(OC)C(C)=C5)=O)=C4C2

InChI key

DUCNNEYLFOQFSW-PMERELPUSA-N

Related Categories

Biochem/physiol Actions

GSK864 helps to decrease cell viability and stimulates apoptosis of tumor cells.[1]
GSK864 is a cell penetrant, potent and selective allosteric inhibitor of isocitrate dehydrogenase 1 (IDH1) that potently inhibits intracellular 2-hydroxyglutarate (2-HG) production in HT-1080 cells. It appears that GSK864 binds to an allosteric binding site and locks WT and mutant IDH1s in a catalytically inactive conformation. GSK864 is a highly bioavailable analog of GSK321. For full characterization details, please visit the GSK864 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for protein targets, visit sigma.com/sgc
GSK864 is a cell penetrant, potent and selective allosteric inhibitor of isocitrate dehydrogenase 1 (IDH1).

Features and Benefits

GSK864 is a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC chemical probes, visit sigma.com/SGC.
This compound is a featured product for Nitric Oxide & Cell Stress research. Click here to discover more featured Nitric Oxide & Cell Stress products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

GSK864 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the GSK864 probe summary on the Chemical Probes Portal website.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cancer-associated IDH1 promotes growth and resistance to targeted therapies in the absence of mutation.
Calvert AE, et al.
Cell Reports, 19(9), 1858-1873 (2017)

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service