Skip to Content
Merck
All Photos(1)

Key Documents

SML3063

Sigma-Aldrich

Pridopidine

≥98% (HPLC)

Synonym(s):

4-[3-(Methylsulfonyl)phenyl]-1-propylpiperidine, ACR 16, ACR16, Huntexil

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€88.40
25 MG
€306.00

€88.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€88.40
25 MG
€306.00

About This Item

Empirical Formula (Hill Notation):
C15H23NO2S
CAS Number:
Molecular Weight:
281.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€88.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCCN1CCC(C2=CC(S(C)(=O)=O)=CC=C2)CC1

InChI

1S/C15H23NO2S/c1-3-9-16-10-7-13(8-11-16)14-5-4-6-15(12-14)19(2,17)18/h4-6,12-13H,3,7-11H2,1-2H3

InChI key

YGKUEOZJFIXDGI-UHFFFAOYSA-N

Biochem/physiol Actions

Pridopidine (ACR16) is orally available, highly selective and potent Sigma-1 Receptor (S1R) agonist that exhibit potent neuroprotective activities. At higher doses pridopidine targets other receptors including dopamine D3, adrenergic α2C and serotoninergic 5-HT1A.
orally available, highly selective and potent Sigma-1 Receptor (S1R) agonist that exhibit potent neuroprotective activities

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service