Skip to Content
Merck
All Photos(1)

Key Documents

M1323

Sigma-Aldrich

Midostaurin hydrate

≥98% (HPLC), solid, kinase inhibitor

Synonym(s):

CGP 41251, N-[(9S,10R,11R,13R)-2,3,10,11,12,13-Hexahydro-10-methoxy-9-methyl-1-oxo-9,13-epoxy-1H,9H-diindolo[1,2,3-gh:3′,2′,1′-lm]pyrrolo[3,4-j][1,7]benzodiazonin-11-yl]-N-methylbenzamide, N-benzoylstaurosporine, PKC-412

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€124.00
5 MG
€469.00

€124.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€124.00
5 MG
€469.00

About This Item

Empirical Formula (Hill Notation):
C35H30N4O4 · xH2O
CAS Number:
Molecular Weight:
570.64 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€124.00


Check Cart for Availability

Request a Bulk Order

Product Name

Midostaurin hydrate, ≥98% (HPLC), solid

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

O.CO[C@@H]1[C@@H](C[C@H]2O[C@]1(C)[n@@H]3c4ccccc4c5c6CNC(=O)c6c7c8ccccc8[n@H]2c7c35)N(C)C(=O)c9ccccc9

InChI

1S/C35H30N4O4.H2O/c1-35-32(42-3)25(37(2)34(41)19-11-5-4-6-12-19)17-26(43-35)38-23-15-9-7-13-20(23)28-29-22(18-36-33(29)40)27-21-14-8-10-16-24(21)39(35)31(27)30(28)38;/h4-16,25-26,32H,17-18H2,1-3H3,(H,36,40);1H2/t25-,26-,32-,35+;/m1./s1

InChI key

YEKDLUZCCDJYAJ-LJAYRLSQSA-N

Biochem/physiol Actions

Midostaurin is an inhibitor of tyrosine kinase, protein kinase C, and VEGF.
Midostaurin is an inhibitor of tyrosine kinase, protein kinase C, and VEGF. Midostaurin inhibits cell growth and phosphorylation of FLT3, STAT5, and ERK. Midostaurin is a potent inhibitor of a spectrum of FLT3 activation loop mutations.

Features and Benefits

This compound is featured on the PKC page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahmoud Hallal et al.
BMC cancer, 21(1), 789-789 (2021-07-10)
Despite the introduction of targeted therapies, most patients with myeloid malignancies will not be cured and progress. Genomics is useful to elucidate the mutational landscape but remains limited in the prediction of therapeutic outcome and identification of targets for resistance.
Jin Kawata et al.
Blood cells, molecules & diseases, 54(2), 206-209 (2014-12-04)
Monocytes and neutrophils are activated during disseminated intravascular coagulation. Tissue factor, the main initiator of coagulation, is expressed by monocytes, while elastase is released by neutrophils. This study investigated tissue factor production by peripheral monocytes after stimulation with human neutrophil
Edmund H Wilkes et al.
Molecular & cellular proteomics : MCP, 16(9), 1694-1704 (2017-07-05)
Cell survival is regulated by a signaling network driven by the activity of protein kinases; however, determining the contribution that each kinase in the network makes to such regulation remains challenging. Here, we report a computational approach that uses mass
Ikuko Omori et al.
Experimental hematology, 52, 56-64 (2017-05-17)
In core-binding factor acute myeloid leukemia (CBF-AML), there have been conflicting reports regarding the status as an unfavorable prognostic factor of mutation in the KIT gene, the significance of which remains unclear. We previously reported that prognoses differ between the
Ee Lin Wong et al.
Nature communications, 10(1), 66-66 (2019-01-10)
Protein-templated fragment ligations have been established as a powerful method for the assembly and detection of optimized protein ligands. Initially developed for reversible ligations, the method has been expanded to irreversible reactions enabling the formation of super-additive fragment combinations. Here

Articles

Protein kinase C (PKC) is an AGC kinase that phosphorylates serine and threonine residues in many target proteins.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service