Skip to Content
Merck
All Photos(1)

Key Documents

SML3062

Sigma-Aldrich

BSJ-04-122-R

≥98% (HPLC)

Synonym(s):

N-(2-((5-chloro-7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)phenyl)propionamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€88.40
25 MG
€285.00

€88.40


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€88.40
25 MG
€285.00

About This Item

Empirical Formula (Hill Notation):
C15H14ClN5O
CAS Number:
Molecular Weight:
315.76
UNSPSC Code:
12352200
NACRES:
NA.77

€88.40


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CNC2=NC=NC(NC3=CC=CC=C3NC(CC)=O)=C21

InChI

1S/C15H14ClN5O/c1-2-12(22)20-10-5-3-4-6-11(10)21-15-13-9(16)7-17-14(13)18-8-19-15/h3-8H,2H2,1H3,(H,20,22)(H2,17,18,19,21)

InChI key

XWBFUHLMYSMDTN-UHFFFAOYSA-N

Biochem/physiol Actions

BSJ-04-122-R is a reversible control for a potent and selective covalent inhibitor of MKK4 and MKK7 BSJ-04-122. BSJ-04-122-R des not inhibit JNK phosphorylation.
reversible control for a potent and selective covalent inhibitor of MKK4 and MKK7 BSJ-04-122

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Jiang et al.
Cell chemical biology, 27(12), 1553-1560 (2020-09-12)
MKK4/7 are kinases that phosphorylate JNKs and regulate the MAPK signaling pathway. Their overexpression has been associated with tumorigenesis and aggressiveness in cancers such as breast, prostate, non-small cell lung, and pediatric leukemia, making them a potential target for inhibitor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service