Skip to Content
Merck
All Photos(1)

Key Documents

SML3018

Sigma-Aldrich

TH1834

≥98% (HPLC)

Synonym(s):

2-(5-(4-((Phenethyl(4-(4-(pyrrolidin-1-ylmethyl)phenoxy)butyl)amino)methyl)phenyl)-2H-tetrazol-2-yl)acetic acid dihydrochloride, 2H-Tetrazole-2-acetic acid, 5-[4-[[(2-phenylethyl)[4-[4-(1-pyrrolidinylmethyl)phenoxy]butyl]amino]methyl]phenyl]-, hydrochloride (1:2), TIP60i

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€125.00
25 MG
€403.00

€125.00


Estimated to ship on19 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€125.00
25 MG
€403.00

About This Item

Empirical Formula (Hill Notation):
C33H40N6O3 · 2HCl
CAS Number:
Molecular Weight:
641.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€125.00


Estimated to ship on19 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(O)CN1N=C(C2=CC=C(CN(CCC3=CC=CC=C3)CCCCOC4=CC=C(CN5CCCC5)C=C4)C=C2)N=N1.[H]Cl.[H]Cl

InChI key

HYXIFSOROZPMMU-UHFFFAOYSA-N

Biochem/physiol Actions

Cell penetrant, potent and selective HAT Tip60 inhibitor
TH1834 is a cell penetrant, potent and selective histone acetyltransferase (HAT) Tip60 inhibitor that interact with active binding pocket of Tip60. TH1834 induces apoptosis and increased unrepaired DNA damage in breast and prostate cancer cell lines but not control MCF10A cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mouhamed Idrissou et al.
Omics : a journal of integrative biology, 24(10), 581-591 (2020-09-23)
Breast cancer is often sporadic due to several factors. Among them, the deregulation of epigenetic proteins may be involved. TIP60 or KAT5 is an acetyltransferase that regulates gene transcription through the chromatin structure. This pleiotropic protein acts in several cellular
James A L Brown et al.
Biochemical Society transactions, 44(4), 979-986 (2016-08-17)
Two opposing enzyme classes regulate fundamental elements of genome maintenance, gene regulation and metabolism, either through addition of an acetyl moiety by histone acetyltransferases (HATs) or its removal by histone de-acetyltransferases (HDAC), and are exciting targets for drug development. Importantly
Chunxia Gao et al.
Scientific reports, 4, 5372-5372 (2014-06-21)
Histone acetylation is required for many aspects of gene regulation, genome maintenance and metabolism and dysfunctional acetylation is implicated in numerous diseases, including cancer. Acetylation is regulated by histone acetyltransferases (HATs) and histone deacetylases and currently, few general HAT inhibitors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service