Skip to Content
Merck
All Photos(1)

Key Documents

208750

Sigma-Aldrich

ALLN

InSolution, ≥95%, Inhibitor of calpain I, calpain II, cathepsin B, and cathepsin L

Synonym(s):

InSolution ALLN, Calpain Inhibitor I, LLNL, MG 101, Proteasome Inhibitor V

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€123.00

€123.00


Estimated to ship on29 May 2025



Select a Size

Change View
5 MG
€123.00

About This Item

Empirical Formula (Hill Notation):
C20H37N3O4
CAS Number:
Molecular Weight:
383.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€123.00


Estimated to ship on29 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

clear colorless

shipped in

wet ice

storage temp.

−20°C

SMILES string

N([C@@H](CC(C)C)C(=O)N[C@@H](CCCC)C=O)C(=O)[C@@H](NC(=O)C)CC(C)C

InChI

1S/C20H37N3O4/c1-7-8-9-16(12-24)22-19(26)18(11-14(4)5)23-20(27)17(10-13(2)3)21-15(6)25/h12-14,16-18H,7-11H2,1-6H3,(H,21,25)(H,22,26)(H,23,27)/t16-,17-,18-/m0/s1

InChI key

FMYKJLXRRQTBOR-BZSNNMDCSA-N

General description

A 10 mM solution of ALLN (Cat. No. 208719) in anhydrous DMSO. Inhibitor of calpain I (Ki = 190 nM), calpain II (Ki = 220 nM), cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 0.5 nM). Inhibits neutral cysteine proteases and proteasome (Ki = 6 µM). Protects against neuronal damage caused by hypoxia and ischemia. Inhibits apoptosis in thymocytes and metamyelocytes. Inhibits cell cycle progression at the G1/S border and metaphase/anaphase in CHO cells by inhibiting cyclin B degradation. Also prevents nitric oxide production by activated macrophages by interfering with transcription of the inducible nitric oxide synthase gene. Inhibits the proteolysis of IkB-α and IkB-β by the ubiquitin-proteasome complex.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
calpain 1
Product does not compete with ATP.
Reversible: no
Target Ki: 190 nM, 220 nM, 150 nM, 500 pM, against calpain I, calpain II, cathepsin B, and cathepsin L, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Flammable (J)

Physical form

A 10 mM (5 mg/1.30 ml) solution of ALLN (Cat. No. 208719) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Milligan, S.A., et al. 1996. Arch. Biochem. Biophys.335, 388.
Griscavage, J.M., et al. 1995. Biochem. Biophys. Res. Commun.215, 721.
Squier, M.K., et al. 1994. J. Cell Physiol.159, 229.
Rami, J., and Kreiglstein, J. 1993. Brain Res.609, 67.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergio Castro-Gonzalez et al.
Retrovirology, 18(1), 33-33 (2021-10-30)
Autophagy plays an important role as a cellular defense mechanism against intracellular pathogens, like viruses. Specifically, autophagy orchestrates the recruitment of specialized cargo, including viral components needed for replication, for lysosomal degradation. In addition to this primary role, the cleavage
Allen C T Teng et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(23), 7165-7170 (2015-06-04)
Phospholamban (PLN) is an effective inhibitor of the sarco(endo)plasmic reticulum Ca(2+) ATPase (SERCA). Here, we examined PLN stability and degradation in primary cultured mouse neonatal cardiomyocytes (CMNCs) and mouse hearts using immunoblotting, molecular imaging, and [(35)S]methionine pulse-chase experiments, together with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service