Skip to Content
Merck
All Photos(1)

Key Documents

SML2981

Sigma-Aldrich

EN219

≥98% (HPLC)

Synonym(s):

1-(3,5-bis(4-Bromophenyl)-4,5-dihydro-1H-pyrazol-1-yl)-2-chloroethanone, 3,5-bis(4-Bromophenyl)-1-(chloroacetyl)-4,5-dihydro-1H-pyrazole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€115.00
25 MG
€372.00

€115.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€115.00
25 MG
€372.00

About This Item

Empirical Formula (Hill Notation):
C17H13Br2ClN2O
CAS Number:
Molecular Weight:
456.56
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€115.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Brc1ccc(cc1)C2N(NC(=C2)c3ccc(cc3)Br)C(=O)CCl

InChI key

PREWPJFURADJST-UHFFFAOYSA-N

Biochem/physiol Actions

EN219 is a cell-permeable chloroacetamide compound that selectively inhibits E3 ligase RNF114-mediated autoubiquitination and p21 ubiquitination. EN219 is shown to covalently interact with RNF114-C8 (IC50 = 470 nM), TUBB1-C201, HSPD1-C442, and HIST1H3A-C97. EN219 is a recruiter of the E3 ubiquitin ligase RNF114 that can be utilized for PROTAC applications.
cell-permeable chloroacetamide compound that selectively inhibits E3 ligase RNF114-mediated autoubiquitination and p21 ubiquitination

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service