Skip to Content
Merck
All Photos(1)

Key Documents

79229

Sigma-Aldrich

N-Boc-1,6-hexanediamine

≥98.0% (NT)

Synonym(s):

N-Boc-1,6-diaminohexane, tert-Butyl-N-(6-aminohexyl)carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€40.30
5 G
€176.00

€40.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€40.30
5 G
€176.00

About This Item

Empirical Formula (Hill Notation):
C11H24N2O2
CAS Number:
Molecular Weight:
216.32
Beilstein:
2219107
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€40.30


Check Cart for Availability

Request a Bulk Order

Assay

≥98.0% (NT)

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.462

density

0.965 g/mL at 20 °C (lit.)

functional group

Boc
amine

SMILES string

NCCCCCCNC(OC(C)(C)C)=O

InChI

1S/C11H24N2O2/c1-11(2,3)15-10(14)13-9-7-5-4-6-8-12/h4-9,12H2,1-3H3,(H,13,14)

InChI key

RVZPDKXEHIRFPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Boc-1,6-hexanediamine can be used as a linear hexyl spacer (C6-spacer) to synthesize:
  • Biodegradable poly(disulfide amine)s for gene delivery.[1]
  • A multifunctional dendrimer for theranostics.[2]
  • Polyamide platinum anti-cancer complexes designed to target cancer specific DNA sequences.[3]
  • Self-assembled monolayers (SAMs) that resist adsorption of proteins.[4]
  • [N-(6-(4-Hydroxy-6-isopropylamino-1,3,5-triazin-2-ylamino)hexyl)-5-hydroxy-1,4-naphthoquinone-3-propionamide] (JUG-HATZ), which can be used in designing electrochemical immunosensors.[5]
Reagent for the introduction of a C6-spacer.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of mixed self-assembled monolayers (SAMs) that resist adsorption of proteins using the reaction of amines with a SAM that presents interchain carboxylic anhydride groups.
Chapman R G, et al.
Langmuir, 16(17), 6927-6936 (2000)
Construction of a well-defined multifunctional dendrimer for theranostics.
Ornelas C, et al.
Organic Letters, 13(5), 976-979 (2011)
Yan Wang et al.
Journal of controlled release : official journal of the Controlled Release Society, 328, 970-984 (2020-09-15)
To avoid poly(ethylene glycol)-related issues of nanomedicines such as accelerated blood clearance, fully N-2-hydroxypropyl methacrylamide (HPMAm)-based polymeric micelles decorated with biotin for drug delivery were designed. To this end, a biotin-functionalized chain transfer agent (CTA), 4-cyano-4-[(dodecylsulfanylthiocarbonyl)-sulfanyl]pentanoic acid (biotin-CDTPA), was synthesized
Design and synthesis of substrates for newborn screening of Maroteaux?Lamy and Morquio A syndromes.
Duffey TA, et al.
Bioorganic & Medicinal Chemistry, 20(20), 5994-5996 (2010)
Synthesis, penetrability and intracellular targeting of fluorescein-tagged peptoids and peptide?peptoid hybrids
Unciti-Broceta A, et al.
Bioorganic & Medicinal Chemistry, 17(3), 959-966 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service