Skip to Content
Merck
All Photos(1)

Key Documents

SML2820

Sigma-Aldrich

SR3335

≥98% (HPLC)

Synonym(s):

ML 176, ML-176, ML176, N-(4-(1,1,1,3,3,3-Hexafluoro-2-hydroxypropan-2-yl)phenyl)thiophene-2-sulfonamide, N-[4-[2,2,2-Trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]phenyl]-2-thiophenesulfonamide, SR 3335, SR-3335

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€85.90
25 MG
€279.00

€85.90


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€85.90
25 MG
€279.00

About This Item

Empirical Formula (Hill Notation):
C13H9F6NO3S2
CAS Number:
Molecular Weight:
405.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€85.90


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)C(O)(C(F)(F)F)c1ccc(cc1)N[S](=O)(=O)c2[s]ccc2

InChI

1S/C13H9F6NO3S2/c14-12(15,16)11(21,13(17,18)19)8-3-5-9(6-4-8)20-25(22,23)10-2-1-7-24-10/h1-7,20-21H

InChI key

LZWUNZRMANFRAO-UHFFFAOYSA-N

Biochem/physiol Actions

SR3335 is a selective partial inverse agonist against the retinoic acid receptor-related orphan receptor ROR-alpha (RORα LBD Ki = 220 nM) that selectively inhibits the constitutive transactivation activity of RORα (IC50 = 480 nM), but not RORβ, RORγ or LXRα, by cell-based reporter assays without affinity toward other RORs. SR3335 effectively suppresses HepG2 cellular RORα target genes expression (5 μM) and improves pyruvate tolerance in DIO mice in vivo (15 mg/kg bid. Ip.) by suppressing gluconeogenesis.
Selective retinoic acid receptor-related receptor RORα partial inverse agonist that improves pyruvate tolerance in DIO mice in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chloé Monnier et al.
Physiological reports, 6(8), e13678-e13678 (2018-04-20)
The RORα-deficient staggerer (sg/sg) mouse is lean and resistant to diet-induced obesity. Its thermogenic activity was shown to be increased not only in brown adipose tissue (BAT), but also in subcutaneous white adipose tissue (WAT) where UCP1 content was enhanced
Ichiaki Ito et al.
Journal of leukocyte biology, 102(6), 1451-1460 (2017-09-28)
Gut microbiota that invades to the defective mucosal barrier is one of the major sources of infectious complications in severely burned hosts. In this study, a role of group 2 innate lymphoid cells (ILC2) and effects of N-{4-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]phenyl}-2-thiophenesulfonamide (SR3335) on
Jun Dai et al.
The Journal of investigative dermatology, 137(12), 2523-2531 (2017-08-05)
The retinoic acid receptor-related orphan receptors RORα and RORγ are critical for the functions of specific subsets of T cells and innate lymphoid cells, which are key drivers of inflammatory disease in barrier tissues. Here, we investigate the anti-inflammatory potential
Yong-Hyun Han et al.
Cell reports, 20(1), 124-135 (2017-07-07)
The regulation of M1/M2 polarization in liver macrophages is closely associated with the progression of nonalcoholic steatohepatitis (NASH); however, the mechanism involved in this process remains unclear. Here, we describe the orphan nuclear receptor retinoic-acid-related orphan receptor α (RORα) as
Charu Rajput et al.
American journal of physiology. Lung cellular and molecular physiology, 312(6), L983-L993 (2017-04-01)
Early-life wheezing-associated respiratory tract infection by rhinovirus (RV) is considered a risk factor for asthma development. We have shown that RV infection of 6-day-old BALB/c mice, but not mature mice, induces an asthmalike phenotype that is associated with an increase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service