Skip to Content
Merck

Skip To

SML2225

RO5263397

≥98% (HPLC), TAAR1 agonist, powder

Synonym(s):

(S)-4-(3-Fluoro-2-methyl-phenyl)-4,5-dihydro-oxazol-2-ylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 90,70
€ 77,09
25 mg
Check Cart for Availability
€ 369,00
€ 313,65

About This Item

Empirical Formula (Hill Notation):
C10H11FN2O
CAS Number:
Molecular Weight:
194.21
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder

€ 77,09

List Price€ 90,70Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

RO5263397, ≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +77 to +88°, c = 1.0 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=N[C@@H](C2=CC=CC(F)=C2C)CO1

InChI

1S/C10H11FN2O/c1-6-7(3-2-4-8(6)11)9-5-14-10(12)13-9/h2-4,9H,5H2,1H3,(H2,12,13)/t9-/m1/s1

InChI key

IOHOUWIYOVWGHV-SECBINFHSA-N

Biochem/physiol Actions

Orally available, potent and selective partial TAAR1 agonist with similar in vivo efficacy as the full agonist RO5256390.
RO5256390 is an orally available partial agonist that targets trace amine-associated receptor 1 (TAAR1) with high affinity (Ki in nM = 0.9/mouse, 4.1/human, 9.1/rat, 24/monkey TARR1), potency (EC50 in nM/relative efficacy with respect to β-phenylethylamine = 1.3/0.59/mouse, 17/0.81/human, 47/0.76/rat, 251/0.85/monkey TARR1-dependent cellular cAMP production), and selectivity (by a 112-receptor/channel/transporter and a 42-enzyme panel), without agonistic activity toward mouse TAAR4-expressing cells even at a high concentration of 30 μM. When applied via oral administration (0.003-1 mg/kg), RO5256390 exhibits similar in vivo efficacy as the full agonist RO5256390 in blocking psychostimulants-induced hyperlocomotion in mice.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1893SML2761SML1990
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige

optical activity

[α]/D +77 to +88°, c = 1.0 in methanol

optical activity

[α]/D 14 to 19°, c = 1 in methanol

optical activity

[α]/D -265 to -295°, c = 0.5 in methanol

optical activity

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters