Skip to Content
Merck
All Photos(1)

Key Documents

SML2664

Sigma-Aldrich

GNF-4877

≥98% (HPLC)

Synonym(s):

(3R)-1-[3-[[[3-Amino-6-[2-fluoro-5-(1-methylethoxy)phenyl]-2-pyrazinyl]carbonyl]amino]-4-pyridinyl]-3-piperidinecarboxylic acid, GNF 4877

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€95.20
25 MG
€305.00

€95.20


Estimated to ship on29 April 2025



Select a Size

Change View
5 MG
€95.20
25 MG
€305.00

About This Item

Empirical Formula (Hill Notation):
C25H27FN6O4
CAS Number:
Molecular Weight:
494.52
UNSPSC Code:
12352200
NACRES:
NA.77

€95.20


Estimated to ship on29 April 2025


Assay

≥98% (HPLC)

form

powder

color

faint yellow to dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=NC=C(C2=CC(OC(C)C)=CC=C2F)N=C1C(NC3=CN=CC=C3N4C[C@H](C(O)=O)CCC4)=O

InChI

1S/C25H27FN6O4/c1-14(2)36-16-5-6-18(26)17(10-16)19-12-29-23(27)22(30-19)24(33)31-20-11-28-8-7-21(20)32-9-3-4-15(13-32)25(34)35/h5-8,10-12,14-15H,3-4,9,13H2,1-2H3,(H2,27,29)(H,31,33)(H,34,35)/t15-/m1/s1

InChI key

UZIATSFXNVOVFE-OAHLLOKOSA-N

Biochem/physiol Actions

GNF-4877 is an orally available dual inhibitor of Dyrk1a and Gsk3b that induces proliferation of functional primary human and rodent β-cells. GNF4877 reduces non-fasting blood glucose levels in a mouse model of T1D.
orally available dual inhibitor of Dyrk1a and Gsk3b that induces proliferation of functional primary human and rodent β-cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Shirakawa et al.
Diabetes, obesity & metabolism, 18 Suppl 1, 71-77 (2016-09-13)
β-Cell dysfunction in type 1 and type 2 diabetes is accompanied by a progressive loss of β-cells, and an understanding of the cellular mechanism(s) that regulate β-cell mass will enable approaches to enhance hormone secretion. It is becoming increasingly recognized
Timothy M Horton et al.
Cell chemical biology, 26(2), 213-222 (2018-12-12)
Diabetes is a hyperglycemic condition characterized by pancreatic β-cell dysfunction and depletion. Whereas methods for monitoring β-cell function in vivo exist, methods to deliver therapeutics to β cells are lacking. We leveraged the rare ability of β cells to concentrate zinc to
Weijun Shen et al.
Nature communications, 6, 8372-8372 (2015-10-27)
Insufficient pancreatic β-cell mass or function results in diabetes mellitus. While significant progress has been made in regulating insulin secretion from β-cells in diabetic patients, no pharmacological agents have been described that increase β-cell replication in humans. Here we report

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service