Skip to Content
Merck
All Photos(1)

Key Documents

SML2617

Sigma-Aldrich

Celiprolol hydrochloride

≥98% (HPLC)

Synonym(s):

Celiprolol hydrochloride, (3-[3-Acetyl-4-(3-t-butylamino-2-hydroxypropoxy)phenyl]-2,1-diethylurea hydrochloride, NSC 324509, REV 5320A, ST 1396, Selecor, Tenoloc

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€89.30
25 MG
€364.00

€89.30


Estimated to ship on23 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€89.30
25 MG
€364.00

About This Item

Empirical Formula (Hill Notation):
C20H33N3O4 · HCl
CAS Number:
Molecular Weight:
415.95
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€89.30


Estimated to ship on23 April 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[Cl-].N(CC)(CC)C(=O)Nc1cc(c(cc1)OCC(O)CNC(C)(C)C)C(=O)C.[H+]

InChI

1S/C20H33N3O4.ClH/c1-7-23(8-2)19(26)22-15-9-10-18(17(11-15)14(3)24)27-13-16(25)12-21-20(4,5)6;/h9-11,16,21,25H,7-8,12-13H2,1-6H3,(H,22,26);1H

InChI key

VKJHTUVLJYWAEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Celiprolol is a selective adrenoceptor modulator that acts not only as a selective β1 receptor antagonist, but also as a β2 receptor partial agonist. Because of its of its β2 agonist activity, celiprolol lacks the typical side effects of the beta-blockers, such as bronchoconstriction, depression of left ventricular function, and peripheral vasoconstriction, making it a good candidate for patients with both cardiovascular and pulmonary disease. It is currently being investigated for treatment of vascular Ehlers-Danlos Syndrome (vEDS).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James J Nawarskas et al.
Cardiology in review, 25(5), 247-253 (2017-07-26)
Celiprolol is a β-blocker with a unique pharmacologic profile: it is a β1-andrenoceptor antagonist with partial β2 agonist activity. Given this combination of effects, celiprolol may be better described as a selective adrenoreceptor modulator. It has antihypertensive and antianginal properties
Lawrence H Kwon et al.
Journal of hypertension, 35(9), 1768-1777 (2017-06-27)
Not only is there a limited number of studies on the effects of vasodilator β-blocker (VBB) therapy on kidney function - specifically, glomerular filtration rate (GFR), serum creatinine (sCr) and proteinuria - but of those that have been reported, the
Jingjing Yu et al.
Journal of pharmaceutical sciences, 106(9), 2312-2325 (2017-04-18)
In recent years, an increasing number of clinical drug-drug interactions (DDIs) have been attributed to inhibition of intestinal organic anion-transporting polypeptides (OATPs); however, only a few of these DDI results were reflected in drug labels. This review aims to provide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service