Skip to Content
Merck
All Photos(1)

Key Documents

SML2614

Sigma-Aldrich

Carboprost tromethamine

≥98% (HPLC)

Synonym(s):

Carboprost trometamol, (15S)-15-methyl prostaglandin F2α tromethamine, (5Z,9a,11a,15E,15S)-9,11,15-Trihydroxy-15-methylprosta-5,13-dien-1-oic acid tris(hydroxymethyl)aminomethane salt, (5Z,9α,11α,13E,15S)-9,11,15-Trihydroxy-15- methylprosta-5,13-dien-1-oic acid with 2-amino-2-(hydroxymethyl)-1,3-propanediol (1:1), Carboprost tromethamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€57.20
25 MG
€299.00

€57.20


Check Cart for Availability


Select a Size

Change View
5 MG
€57.20
25 MG
€299.00

About This Item

Empirical Formula (Hill Notation):
C25H47NO8
CAS Number:
Molecular Weight:
489.64
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.77

€57.20


Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 18 to 24°, c = 1.0 in chloroform-d

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

NC(CO)(CO)CO.O[C@H]1[C@H](C([C@H](C1)O)\C=C/[C@@](O)(CCCCC)C)C\C=C\CCCC(=O)O

InChI

1S/C21H36O5.C4H11NO3/c1-3-4-9-13-21(2,26)14-12-17-16(18(22)15-19(17)23)10-7-5-6-8-11-20(24)25;5-4(1-6,2-7)3-8/h5,7,12,14,16-19,22-23,26H,3-4,6,8-11,13,15H2,1-2H3,(H,24,25);6-8H,1-3,5H2/b7-5+,14-12-;/t16-,17?,18+,19-,21-;/m0./s1

InChI key

UMMADZJLZAPZAW-QFXCEMDRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Carboprost tromethamine is a metabolically stable synthetic analog of prostaglandin F2α. It is used clinically to treat severe postpartum bleeding and for the termination of pregnancy in the second trimester.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service