Skip to Content
Merck
All Photos(1)

Key Documents

SML2560

Sigma-Aldrich

OB-1

≥98% (HPLC)

Synonym(s):

Ethyl 5-(1-methyl-4-nitro-1H-imidazol-5-yloxy)-2-phenylbenzofuran-3-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€71.60
25 MG
€231.00

€71.60


Check Cart for Availability


Select a Size

Change View
5 MG
€71.60
25 MG
€231.00

About This Item

Empirical Formula (Hill Notation):
C21H17N3O6
CAS Number:
Molecular Weight:
407.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€71.60


Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

color

white to very dark brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1nc[n](c1Oc2cc3c([o]c(c3C(=O)OCC)c4ccccc4)cc2)C

InChI key

UJHGRDVYTTVJFD-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibitor of Stomatin-like protein-3 (STOML3) oligomerization
OB-1 is an inhibitor of the oligomerization of Stomatin-like protein-3 (STOML3), which is a modulator of the mechanosensitive ion channel Piezo2. STOML3 requires oligomerization for its role as an endogenous regulator of the sensitivity of mechanosensitive Piezo ion channels in sensory neurons. By inhibiting this activity, OB-1 has been shown to reduce touch-evoked pain behavior in two mouse models of neuropathic pain.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service