Skip to Content
Merck
All Photos(1)

Key Documents

SML0060

Sigma-Aldrich

NB001

≥98% (HPLC)

Synonym(s):

5-[[2-(6-Amino-9H-purin-9-yl)ethyl]amino]-1-pentanol; HTS 09836

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20N6O
CAS Number:
Molecular Weight:
264.33
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: ≥24 mg/mL

storage temp.

−20°C

SMILES string

Nc1ncnc2n(CCNCCCCCO)cnc12

InChI

1S/C12H20N6O/c13-11-10-12(16-8-15-11)18(9-17-10)6-5-14-4-2-1-3-7-19/h8-9,14,19H,1-7H2,(H2,13,15,16)

InChI key

CVPTTZZCRDVGSU-UHFFFAOYSA-N

Application

NB001 may be used in adenylyl cyclase 1-mediated cell signaling studies.

Biochem/physiol Actions

NB001, originally believed to be a selective inhibitor of adenylyl cyclase 1 (AC1), new evidence suggests NB001 does not interact with AC1 directly. NB001 may reduce cAMP accumulation in cells through an unknown mechanism that is dependent on AC1.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Adenylyl cyclases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth Stanley Shepherd et al.
Nature, 557(7705), 434-438 (2018-05-11)
The dense microbial ecosystem in the gut is intimately connected to numerous facets of human biology, and manipulation of the gut microbiota has broad implications for human health. In the absence of profound perturbation, the bacterial strains that reside within
Kelly E Bosse et al.
The Journal of pharmacology and experimental therapeutics, 363(2), 148-155 (2017-08-26)
Neuroadaptive responses to chronic ethanol, such as behavioral sensitization, are associated with
Sung-Chul Lim et al.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology, 21(4), 397-405 (2017-07-15)
MDL-12330A is a widely used adenylyl cyclase (AC) inhibitor that blocks AC/cAMP signaling. In this study, we demonstrated a novel antitumor activity of this drug in gastric carcinoma (GC) cell lines. In these GC cells, MDL-12330A reduced cell viability and
Ryan B Griggs et al.
Neurobiology of disease, 127, 76-86 (2019-02-27)
Painful diabetic neuropathy (PDN) is a devastating neurological complication of diabetes. Methylglyoxal (MG) is a reactive metabolite whose elevation in the plasma corresponds to PDN in patients and pain-like behavior in rodent models of type 1 and type 2 diabetes.
Shui-Bing Liu et al.
Journal of neurochemistry, 153(2), 252-263 (2019-10-31)
Cortical areas including the anterior cingulate cortex (ACC) play critical roles in different types of chronic pain. Most of previous studies focus on the sensory inputs from somatic areas, and less information about plastic changes in the cortex for visceral

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service