Skip to Content
Merck
All Photos(1)

Key Documents

SML2555

Sigma-Aldrich

BIO-0919278 racemate

≥98% (HPLC)

Synonym(s):

BIO-0702697 racemate, BIO0702697 racemate, BIO0919278 racemate, Compound 702697 racemate, Compound 919278 racemate, N-(1H-Benzo[d]imidazol-2-yl)-2-(1-oxoisoindolin-2-yl)propenamide, N-1H-Benzimidazol-2-yl-1,3-dihydro-a-methyl-1-oxo-2H-isoindole-2-acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€71.60
25 MG
€231.00

€71.60


Estimated to ship on30 May 2025



Select a Size

Change View
5 MG
€71.60
25 MG
€231.00

About This Item

Empirical Formula (Hill Notation):
C18H16N4O2
CAS Number:
Molecular Weight:
320.35
UNSPSC Code:
12352200
NACRES:
NA.77

€71.60


Estimated to ship on30 May 2025


Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1c2c([nH][c]1=NC(=O)C(N3Cc4c(cccc4)C3=O)C)cccc2

InChI key

PWSSISUOJZBZFT-UHFFFAOYSA-N

Biochem/physiol Actions

BIO-0919278 is a highly selective cyclin-dependent kinase CDK12 inhibitor (Kd = 5.6 μM) with little or no potency against CDK1-7, CDK9, CDK14, CDK16-18 or 344 other kinases (<35% inhibition at 1 μM). BIO-0919278 reduces CDK12-mediated Pol II CTD Ser2 phosphorylation (by 57% at 1 μM; U-2 OS cells) without affecting CDK12-independent Pol II CTD Ser5 phosphorylation, and selectively targets noncanonical NF-κB activation (4-hr 20 ng/mLTWEAK-induced p52 nuclear translocation/NIK mRNA production IC50 = 167/320 nM; U-2 OS cells) without affecting canonical NF-κB activation (TNF-α-stimulated p65 (RelA) nuclear translocation IC50 >30 μM).
Highly selective CDK12 inhibitor that disrupts noncanonical, but not canonical, NF-κB activation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service