Skip to Content
Merck
All Photos(1)

Key Documents

SML2465

Sigma-Aldrich

CY-09

≥98% (HPLC)

Synonym(s):

4-[[4-Oxo-2-thioxo-3-[[3-(trifluoromethyl)phenyl]methyl]-5-thiazolidinylidene]methyl]benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€87.30
25 MG
€355.00

€87.30


Estimated to ship on05 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€87.30
25 MG
€355.00

About This Item

Empirical Formula (Hill Notation):
C19H12F3NO3S2
CAS Number:
Molecular Weight:
423.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€87.30


Estimated to ship on05 April 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

faint yellow to dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C(C=C1)=CC=C1/C=C(SC2=S)\C(N2CC3=CC=CC(C(F)(F)F)=C3)=O)=O

InChI

1S/C19H12F3NO3S2/c20-19(21,22)14-3-1-2-12(8-14)10-23-16(24)15(28-18(23)27)9-11-4-6-13(7-5-11)17(25)26/h1-9H,10H2,(H,25,26)/b15-9+

InChI key

DJTINRHPPGAPLD-OQLLNIDSSA-N

Biochem/physiol Actions

CY-09 is an NLRP3 inflammasone inhibitor. CY-09 binds directly to the ATP-binding motif of NLRP3 NACHT domain and inhibits its ATPase activity, blocking NLRP3 inflammasome assembly and activation. In mouse models CY-09 showed therapeutic effects in type 2 diabetes and cryopyrin-associated auto-inflammatory syndrome (CAPS), which is caused by gain-of-function mutations of NLRP3.
NLRP3 inflammasome inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianlin Qiao et al.
Haematologica, 103(9), 1568-1576 (2018-05-26)
In addition to their hemostatic function, platelets play an important role in regulating the inflammatory response. The platelet NLRP3 inflammasome not only promotes interleukin-1β secretion, but was also found to be upregulated during platelet activation and thrombus formation in vitro
Matthew S J Mangan et al.
Nature reviews. Drug discovery, 17(8), 588-606 (2018-07-22)
Danger signals are a hallmark of many common inflammatory diseases, and these stimuli can function to activate the cytosolic innate immune signalling receptor NLRP3 (NOD-, LRR- and pyrin domain-containing 3). Once activated, NLRP3 nucleates the assembly of an inflammasome, leading
Hua Jiang et al.
The Journal of experimental medicine, 214(11), 3219-3238 (2017-10-13)
The NLRP3 inflammasome has been implicated in the pathogenesis of a wide variety of human diseases. A few compounds have been developed to inhibit NLRP3 inflammasome activation, but compounds directly and specifically targeting NLRP3 are still not available, so it
Zihao Li et al.
Cell death discovery, 7(1), 346-346 (2021-11-12)
Instability and excessive use of the knee joint can cause osteoarthritis (OA). Reasonable exercise can enhance the stability of the knee joint and prevent and relieve the occurrence and development of OA. As a key switch for inflammation, P2X purinoceptor
Andrea Mencarelli et al.
Nature immunology, 25(4), 693-702 (2024-03-15)
The inflammasome components NLRP3 and ASC are cytosolic proteins, which upon sensing endotoxins or danger cues, form multimeric complexes to process interleukin (IL)-1β for secretion. Here we found that antigen (Ag)-triggered degranulation of IgE-sensitized mast cells (MCs) was mediated by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service