Skip to Content
Merck
All Photos(1)

Key Documents

SML2118

Sigma-Aldrich

HA15

≥98% (HPLC)

Synonym(s):

N-[4-[3-[[[5-(Dimethylamino)-1-naphthalenyl]sulfonyl]amino]phenyl]-2-thiazolyl]-acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€105.00
25 MG
€427.00

€105.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€105.00
25 MG
€427.00

About This Item

Empirical Formula (Hill Notation):
C23H22N4O3S2
CAS Number:
Molecular Weight:
466.58
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€105.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(NC1=NC(C2=CC=CC(NS(C3=C(C=CC=C4N(C)C)C4=CC=C3)(=O)=O)=C2)=CS1)=O

InChI

1S/C23H22N4O3S2/c1-15(28)24-23-25-20(14-31-23)16-7-4-8-17(13-16)26-32(29,30)22-12-6-9-18-19(22)10-5-11-21(18)27(2)3/h4-14,26H,1-3H3,(H,24,25,28)

InChI key

LBSMEKVVMYSTIH-UHFFFAOYSA-N

Biochem/physiol Actions

HA15 is a selective ligand of BiP/GRP78/HSPA5 chaperone that increases ER stress, leading to autophagic and apoptotic cell death in varies cancer cell lines including melanoma cells. HA15 inhibits inhibit the ATPase activity of BiP. HA15 inhibits tumor development in mice injected with A375 melanoma cells.
selective ligand of BiP/GRP78/HSPA5 chaperone that increases ER stress, leading to autophagic and apoptotic cell death in varies cancer cell lines

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaël Cerezo et al.
Autophagy, 13(1), 216-217 (2016-10-30)
Treatment of melanoma has significantly advanced over the last decade, with the development of targeted therapies against the MAPK pathway and immunotherapies to reactivate antitumor immunity. Unfortunately, currently more than 50% of patients are in treatment failure. Thus, identification of
Michaël Cerezo et al.
Cancer cell, 29(6), 805-819 (2016-05-31)
We have discovered and developed a series of molecules (thiazole benzenesulfonamides). HA15, the lead compound of this series, displayed anti-cancerous activity on all melanoma cells tested, including cells isolated from patients and cells that developed resistance to BRAF inhibitors. Our
Rossella Benedetti et al.
Biomedicines, 9(9) (2021-09-29)
Polyphenols have been shown to possess several beneficial properties, including properties involved in the prevention or treatment of cancer. Among these polyphenols, a leading role is played by dihydroxyphenylethanol (DPE), the most powerful antioxidant compound contained in the olive oil.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service